MilliporeSigma
HomeLiquid Formulation StrategiesUse of PVA to Overcome Challenges in Ophthalmic Formulations
How the Use of PVA Can Help Overcome Challenges in Ophthalmic Formulations
Successful ophthalmic drug formulations rely on the selection of the correct polymer to ensure the desired characteristics such as retention in the eye cavity and solubility. One example of such an excipient is polyvinyl alcohol (PVA) which offers many advantages for ophthalmic drug formulations including eye drops, suspensions, and gels. It is a biocompatible synthetic polymer produced by the polymerization of vinyl acetate and partial hydrolysis of the resulting esterified polymer. PVA is recognized as safe (GRAS) by the US Food and Drug Administration (FDA) , does not have any immunogenic effects, and its long-term use has been demonstrated in many different formulations including oral, topical, and ophthalmic.
Section Overview
Advantages of PVA for Ophthalmic Formulation
PVA offers many advantages for ophthalmic formations including eye drops and hydrogels. It is water soluble and has a narrow range of viscosity and a high degree of swelling, offering the precise viscosity needed for formulations to remain in the eye cavity. Scientific evidence and long-term use confirm the physiological compatibility and safety of PVA, while a variety of grades allows selection of the exact properties needed for specific formulations.
Its physicochemical properties make PVA especially well suitable for the application in ophthalmic drug delivery systems including:
  • The ability to form a transparent solution which is essential for medications administered to the eye
  • High adhesion and high correlation properties, which enable retention in the eye cavity
  • Suitability for lubricating eye drops
  • The ability to act as an inhibitor of crystallization which helps retain solubility of the API throughout storage of the dosage form
Selecting the Hydrolysis Grade of PVA Based on Viscosity
As a synthetic polymer, PVA offers batch-to-batch consistency and is available in different grades defined by viscosity, hydrolysis and the microbial load (total aerobic bacteria, total yeast and total mold count), which should be specified by the manufacturer. These grades come with different physicochemical properties, making them suitable for different applications.
Lower viscosity grades of PVA can be used for lubricant activity, to enhance API solubility and as inhibitors of crystallization. If the formulation is a suspension or gel, a high viscosity grade PVA is more suitable and can be used as a thickener or viscosity enhancer. Different hydrolysis grades of PVA, which refer to the amount of residual, unhydrolyzed acetate groups within the polymer chain, also affect the polymer performance. Higher hydrolysis grades improve tensile strength of the hydrogel and provide a stronger gel scaffold through H-bonds while lower hydrolysis grades might be a better choice for drug delivery of poorly water-soluble APIs. However, since some pharmacopoeias restrict the hydrolysis grade variation, there is limited flexibility in this aspect.
Another important consideration when using PVA is the possible presence of crotonaldehyde, which is a strong eye irritant and a process impurity generated during synthesis. While there is no information in the pharmacopeia regarding the limit of this impurity, given its toxic nature, it is important that this impurity content is known and controlled. Due to stringent regulatory requirements in this segment, a multi-compendial product and regulatory and documentation support by the manufacturer is essential to facilitate regulatory submission. We specify low crotonaldehyde limits for PVAs.
PVA Solution Preparation for Ophthalmic Applications
Table 1 compares the preparation, appearance, and presence of foam and particles in solutions of three different polymers commonly used in ophthalmic preparations: PVA, and semi-synthetic options such as hydroxypropylmethylcellulose (HPMC) and carboxymethyl cellulose (CMC). While a higher temperature is recommended for PVA dissolution, the result will be a clear, particle free solution without foam. HPMC and CMC can be dissolved at room temperature, but it may be challenging to obtain particle-free solutions, and foaming was observed during HPMC solution preparation.
Parameters
1.4% PVA Solution
1.0% HPMC Solution
0.5% CMC Solution
Preparation Time
20-35 min
30-45 min
25-30 min
Temperature
90-95 °C
Room temperature
Room temperature
Appearance
Clear
Clear
Little hazy
Foaming
No
Yes
No
Particles
No
Yes
Yes
Table 1.Comparison of solubilization parameters for three polymers commonly used in ophthalmic formulations.
Flasks containing PVA, CMC, and HPMC individually after steam sterilization.
Figure 1.Comparison of CMC, HPMC and PVA following steam sterilization.
Sterilization Methods Affect PVA, HPMC, and CMC Polymers Differently
Different sterilization methods can be used with polymer solutions including steam and filtration, and each affects attributes of the polymer in different ways.
Steam Sterilization
When PVA, HPMC, and CMC are sterilized at 121°C for 15 minutes at 15 PSI, only the PVA solution remains clear and transparent (Figure 1). The CMC solution becomes cloudy while a white colloidal appearance was observed with the HPMC solution turns white colloidal nature which became transparent once cooled to room temperature. PVA and HPMC solutions remain similar following sterilization times of 15, 20, and 25 minutes while the viscosity of CMC decreases significantly after the first 15 minutes of steam sterilization indicating that some degradation might be occurring.
Sterile FIltration
Sterile filtration can also be used to achieve a sterile formulation as demonstrated by the results of a study comparing polymers. The study also found that scale-up is facilitated with use of PVA. Filtration of a PVA 4-88 solution was possible through both 0.2 µm polyvinylidene fluoride (PVDF) and polyethylsulfone (PES) membranes. Of these two options, the PES membrane provided better process economics with a higher Vmax and mean flux. The CMC solution could not be filtered using this method due to high viscosity. In the case of HPMC, filtration was feasible, but the filter size required to process this batch was much larger than for the PVA. Scale up and commercial production are thus more viable with PVA, without any challenge to sterile filtration, as compared to HPMC or CMC.
Excipient Compatibility
A final consideration when selecting an excipient is compatibility with other excipients and the API contained within the final ophthalmic formulation; other excipients may include preservatives, inorganic salts to maintain osmolarity, and buffers. Inorganic salts do not alter the viscosity of PVA, indicating compatibility. In contrast, when CMC comes into contact with the inorganic salts, the viscosity was drastically changed. There is also no change in the viscosity of a PVA solution across a pH range of 5.5 to 8.5 which is the typically preferred pH range for ophthalmic preparations. PVA does precipitate completely when used in ophthalmic formulations containing boric acid. This is one of the very few limitations of PVA.
Advantages of PVA for Ophthalmic Preparations
PVA meets all the essential requirements to be used in ophthalmic preparations and offers important benefits compared to HPMC and CMC (Table 2).
Parameter
PVA
HPMC
CMC
Origin of Source
Synthetic
Semi-synthetic
Semi-synthetic
Viscosity Range / Variation
85-115%
80-120% (low visc)
70-130% (high visc)
>50% variation
Microbial Load
Minimal
High
High
Solution Preparation
Heating at 90-95˚C
Room temperature but foam formation
Room temperature
Ease of Sterile Filtration
Yes
Yes
No
Change in CQA After Filtration
No
No
Not applicable
Steam Sterilization
Yes
Yes
Yes (change in viscosity)
Impact of pH
No
No
No
Impact of Ions
No
No
Yes
Table 2.Comparison of polymers used in ophthalmic formulations shows the clear advantage of PVA.
As they are semi-synthetic in nature, HPMC and CMC have a relatively large range in viscosity compared to PVA and can be expected to have a higher microbial load. While PVA requires a higher temperature for solution preparation, foam and particles have been observed in HPMC and CMC solutions. PVA and HPMC can be sterilized using steam sterilization or sterile filtration with no changes in viscosity. Changes in pH do not affect the PVA solution and this polymer was shown to be compatible with a variety of inorganic salts.
In summary, while several polymers are available for use in ophthalmic formulations, PVA offers important advantages. It is best suited to overcome many formulation challenges and is a viable alternative to HPMC and CMC for ophthalmic formulations, which is why it is an important part of the formulator toolbox for ophthalmics.
Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
References
1.
Polyvinyl alcohol: Revival of a long lost polymer. [Internet]. Kasselkus A, Weiskircher-Hildebrandt E, Schornick E, Bauer F, Zhen M. Available from: https://www.emdmillipore.com/Web-US-Site/en_CA/-/USD/ShowDocument-Pronet?id=202009.075