Skip to Content
Merck

Skip To

214 BLCL Human Blood, ECACC 94031663

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41106514
eCl@ss:
32160703
Biological source:
human blood
Technique(s):
cell culture | mammalian: suitable
Ethnicity:
Caucasian
Gender:
female
Growth mode:
Suspension
Morphology:
Lymphoblastoid
Sign in to see your pricing
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

214 BLCL human cell line, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

human blood

manufacturer/tradename

ECACC

packaging

tube of 5 μg 94031663-DNA-5UG, pkg of vial of cells 94031663-1VL

ethnicity

Caucasian

gender

female

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

-140 to -196°C

Biochem/physiol Actions

Not specified
This cell line is an Epstein-Barr Virus (EBV) transformed lymphoblastoid line and is part of the Human Leukocyte Antigen (HLA) Typed Collection maintained by the European Collection of Cell Cultures (ECACC). HLA-Typing Data is available for this cell line.

Preparation Note

RPMI 1640 (R0883) + 2mM Glutamine (G7513) + 20% Foetal Bovine Serum (FBS) (F2442). Once a growing culture is established the FBS concentration can be reduced to 10%.
Maintain cultures between 3x105 - 2x106 cells/ml, 5% CO2, 37 °C. Upon resuscitation remove cryoprotectant - centrifuge at 300 x g for 5 minutes and re-suspend the cell pellet in fresh culture medium.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
HLA A: 24 26
HLA C: 6 0
HLA B: 13 41
HLA DR: 12 7
HLA DQ: 3 2
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). Only the genomic DNA extracted from the cell line is available for purchase by commercial organizations, and not the cell line itself. No licensing agreement is required when the DNA extracted from this cell line are used for internal research purposes only. Frozen cell cultures can be supplied only to academic and not-for-profit organisations. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding other authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.

Disclaimer

This cell line has restrictions and can only be supplied to academic and not-for-profit organisations. Nucleic acids can be supplied to all researchers including those from commercial organisations.
This product is regulated in France when intended for scientific purposes, including import, export, and conservation activities (Articles L.1211-1, paragraph 2, and L.1243-4 of the French Public Health Code). The purchaser (i.e. end user) is required to obtain an import authorization from the France Ministry responsible for Research (as referenced in Article L.1245-5-1 II of the French Public Health Code) or a conservation authorization from the same ministry (as referenced in Article L.1243-4 of the French Public Health Code). By placing an order for this product, you confirm that you have obtained the appropriate import or conservation authorization for the product. For more information regarding the use of human biological samples for research purposes in France, please contact the relevant French authorities.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CB_94031654CB_94031639CB_94031651
description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

94031639, IHW Number: SLE, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

ethnicity

Caucasian

ethnicity

Caucasian

ethnicity

Caucasian

ethnicity

Caucasian

gender

female

gender

female

gender

female

gender

male

karyotype

Not specified

karyotype

Not specified

karyotype

Not specified

karyotype

-

packaging

pkg of vial of cells 94031663-1VL, tube of 5 μg 94031663-DNA-5UG

packaging

tube of 5 μg 94031654-DNA-5UG, pkg of vial of cells 94031654-1VL

packaging

tube of 5 μg 94031639-DNA-5UG, pkg of vial of cells 94031639-1VL

packaging

tube of 5 μg 94031651-DNA-5UG, pkg of vial of cells 94031651-1VL

storage temp.

-140 to -196°C

storage temp.

-140 to -196°C

storage temp.

-140 to -196°C

storage temp.

-140 to -196°C

manufacturer/tradename

ECACC

manufacturer/tradename

ECACC

manufacturer/tradename

ECACC

manufacturer/tradename

ECACC




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters