Merck
Resveratrol attenuates HMGB1 signaling and inflammation in house dust mite-induced atopic dermatitis in mice.
International immunopharmacology (2014-12-04)
Vengadeshprabhu Karuppagounder, Somasundaram Arumugam, Rajarajan A Thandavarayan, Vigneshwaran Pitchaimani, Remya Sreedhar, Rejina Afrin, Meilei Harima, Hiroshi Suzuki, Mayumi Nomoto, Shizuka Miyashita, Kenji Suzuki, Kenichi Watanabe
RESUMEN
Resveratrol is a polyphenol abundantly found in red grape skin and is effective against antiaging and anti-inflammation associated with immune responses. In this study, we have investigated the effect of resveratrol on skin lesion, high mobility group box (HMGB)1 and inflammation pathway in an atopic dermatitis (AD) mouse model. AD-like lesion was induced by the application of house dust mite extract to the dorsal skin of NC/Nga mouse. After AD induction, resveratrol (20 mg/kg, p.o.) was administered daily for 2 weeks. We evaluated dermatitis severity, histopathological changes, serum levels of T helper (Th) cytokines (interferon (IFN)γ, interleukin (IL)-4) and changes in protein expression by Western blotting for HMGB1, receptor for advanced glycation end products (RAGE), toll like receptor (TLR)4, nuclear factor (NF)κB, phosphatidylinositide 3-kinase (PI3K), extracellular signal-regulated kinase (ERK)1/2, cyclooxygenase (COX)2, tumor necrosis factor (TNF)α, IL-1β, IL-2Rα and other inflammatory markers in the skin of AD mice. Treatment of resveratrol inhibited the development of the AD-like skin lesions. Histological analysis showed that resveratrol inhibited hypertrophy, intracellular edema, mast cells and infiltration of inflammatory cells. Furthermore, resveratrol treatment down-regulated HMGB1, RAGE, p-NFκB, p-PI3K, p-ERK1/2, COX2, TNFα, IL-1β, IL-2Rα, IFNγ and IL-4. Considering all these findings together, the HMGB1 pathway might be a potential therapeutic target in skin inflammation, and resveratrol treatment could have beneficial effects on AD by modulating the HMGB1 protein expression.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
2- Mercaptoetanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2- Mercaptoetanol, ≥99.0%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
2- Mercaptoetanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Cloruro de sodio, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
Supelco
Cloruro de sodio, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cloruro de sodio, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cloruro de sodio, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Cloruro de sodio, tested according to Ph. Eur.
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%