Merck
Targeting liver cancer via ASGP receptor using 5-FU-loaded surface-modified PLGA nanoparticles.
Journal of microencapsulation (2014-04-05)
Ruchi Dangi, Pooja Hurkat, Ankit Jain, Satish Shilpi, Ashish Jain, Arvind Gulbake, Sanjay K Jain
RESUMEN
Liver cancer is widespread liver malignancy in the world, for an estimated one million deaths annually. In present work, lactobionic acid conjugated PLGA nanoparticles (LDNPs) bearing 5-Fluorouracil (5-FU) were developed for targeted delivery to hepatocellular carcinoma. Lactobionic acid conjugated PLGA was used to prepare LDNPs using modified emulsion diffusion method. They were characterised for particle morphology, particle size (below 150 nm), zeta potential and polydispersity index (PDI ∼0.35), entrapment efficiency (∼60.23%), and cumulative percent drug release. LDNPs in ex-vivo cell line studies on human cancer cell line HepG2 exhibited significantly higher cytotoxicity compared to 5-FU and DNPs (unconjugated PLGA NPs) with growth inhibition 50% (GI50) of 66.7 µg/mL, 50.2 µg/mL and 35.5 µg/mL, respectively. In vivo studies exhibited higher drug concentration about 37.52 ± 0.68% in liver as compared to other organs and plasma. Thus, LDNPs showed high drug loading, specificity, biocompatibility and efficacy in treatment of liver cancer.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dimetilsulfóxido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimetilsulfóxido, Molecular Biology
Sigma-Aldrich
Dimetilsulfóxido, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimetilsulfóxido, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimetilsulfóxido, anhydrous, ≥99.9%
Sigma-Aldrich
Dimetilsulfóxido, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimetilsulfóxido, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
D-(+)-Galactosa, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Lactobionic acid, 97% (TLC)
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimetilsulfóxido, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Galactosa, ≥99% (HPLC)
Sigma-Aldrich
Dimetilsulfóxido, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimetilsulfóxido, PCR Reagent
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
D-(+)-Galactosa, ≥98% (HPLC)
Sigma-Aldrich
Dimetilsulfóxido, puriss. p.a., dried, ≤0.02% water
Supelco
D-(+)-Galactosa, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)
Sigma-Aldrich
N,N′-Dicyclohexylurea, 98%
Supelco
Dimetilsulfóxido, analytical standard
Millipore
D-(+)-Galactosa, suitable for microbiology, ≥99.0%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Ethylenediamine, BioXtra
Supelco
Dimetilsulfóxido, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimetilsulfóxido, United States Pharmacopeia (USP) Reference Standard
USP
Galactose, United States Pharmacopeia (USP) Reference Standard