Merck
  • ApoG2 inhibits the antiapoptotic protein, Mcl‑1, and induces mitochondria‑dependent apoptosis in human colorectal cancer cells.
ApoG2 inhibits the antiapoptotic protein, Mcl‑1, and induces mitochondria‑dependent apoptosis in human colorectal cancer cells.
Molecular medicine reports (2015-09-10)
Tianxiao Li, Gang Yuan, Lin Zhang, Lijun Ye, Shuxia Li, Yuhua Fan, Jian Sun
RESUMEN
Colorectal cancer (CRC) is a worldwide malignancy of high incidence and mortality. At present, there is a lack of effective drugs against CRC. The B‑cell leukemia/lymphoma 2 (Bcl‑2) protein family members are considered to be closely associated with tumorigenesis and the chemoresistance of CRC. As a novel gossypol derivative targeting antiapoptotic proteins of the Bcl‑2 family, apogossypolone (ApoG2) exhibits antitumor properties in various cancer types, although its effects against CRC remain to be fully elucidated. In the present study, the cytotoxicity of ApoG2 in vitro on CRC cells was investigated, with the aim of elucidating the underlying mechanism. Using an MTT assay, ApoG2 was revealed to inhibit the growth of the HT29, SW480 and HCT116 CRC cell lines in a dose‑ and a time‑dependent manner. Hoechst staining revealed that ApoG2 induced CRC cell apoptosis, marked by morphological changes, including cell shrinkage and nuclear fragmentation. Flow cytometric analysis also detected a higher apoptotic ratio following treatment with ApoG2. The ratio was dependent upon the concentration of ApoG2, which the cells were exposed to, and the duration of the exposure. Western blot analysis and immunoprecipitation experiments revealed that ApoG2 treatment led to the downregulation of the protein expression of Mcl‑1, and the interruption of the binding of Mcl‑1 to the protein Bax. Furthermore, treatment with ApoG2 led to the release of cytochrome c into the cytoplasm and the activation of caspases 3 and 7. The present study revealed that ApoG2 inhibited the proliferation of the CRC cell lines through mitochondrial signaling pathway‑dependent apoptosis, which may be associated with the disruption of the function of the Mcl‑1 protein by ApoG2.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dimetilsulfóxido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimetilsulfóxido, Molecular Biology
Sigma-Aldrich
Dimetilsulfóxido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimetilsulfóxido, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimetilsulfóxido, anhydrous, ≥99.9%
Sigma-Aldrich
Dodecilsulfatosódico, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Dimetilsulfóxido, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dodecilsulfatosódico, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Dimetilsulfóxido, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Azul de tiazolil Bromuro de tetrazolio, 98%
Sigma-Aldrich
Dodecilsulfatosódico, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Yoduro de propidio, ≥94.0% (HPLC)
Sigma-Aldrich
Azul de tiazolil Bromuro de tetrazolio, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dodecilsulfatosódico, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Dimetilsulfóxido, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dodecilsulfatosódico, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Supelco
Dodecilsulfatosódico, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Dimetilsulfóxido, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dodecilsulfatosódico, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Dimetilsulfóxido, PCR Reagent
Sigma-Aldrich
Dodecilsulfatosódico, ≥98.0% (GC)
Sigma-Aldrich
Dodecilsulfatosódico, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Dimetilsulfóxido, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dodecilsulfatosódico, ≥90% ((Assay))
Sigma-Aldrich
Dodecilsulfatosódico, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O