Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
10 mg | Versand in 21 Tagen - ab Aldrich Partner. | € 381,20 |
50 mg | Versand in 21 Tagen - ab Aldrich Partner. | € 1.020,84 |
100 mg | Versand in 21 Tagen - ab Aldrich Partner. | € 1.378,04 |
Über diesen Artikel
Lieferanten-SKU: AA00C2A0-10MG
€ 381,20
Inchi Code
1S/C53H100O6/c1-4-7-10-13-16-19-22-25-26-29-32-35-38-41-44-47-53(56)59-50(48-57-51(54)45-42-39-36-33-30-27-23-20-17-14-11-8-5-2)49-58-52(55)46-43-40-37-34-31-28-24-21-18-15-12-9-6-3/h25-26,50H,4-24,27-49H2,1-3H3/b26-25+
Inchi Key
FDCOHGHEADZEGF-OCEACIFDSA-N
Storage temp.
2~8C
Purity
99%
Country of Origin
US
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description - | description ≥99% | description - | description - |
| Purity 99% | Purity - | Purity 97% | Purity 99% |
| Inchi Key FDCOHGHEADZEGF-OCEACIFDSA-N | Inchi Key - | Inchi Key - | Inchi Key - |
| Storage temp. 2~8C | Storage temp. - | Storage temp. Inert atmosphere, store in freezer, under -20C | Storage temp. - |
| Country of Origin US | Country of Origin - | Country of Origin CN | Country of Origin US |
| Inchi Code 1S/C53H100O6/c1-4-7-10-13-16-19-22-25-26-29-32-35-38-41-44-47-53(56)59-50(48-57-51(54)45-42-39-36-33-30-27-23-20-17-14-11-8-5-2)49-58-52(55)46-43-40-37-34-31-28-24-21-18-15-12-9-6-3/h25-26,50H,4-24,27-49H2,1-3H3/b26-25+ | Inchi Code - | Inchi Code 1S/C53H100O6/c1-4-7-10-13-16-19-22-25-26-29-31-34-37-40-43-46-52(55)58-49-50(59-53(56)47-44-41-38-35-32-28-24-21-18-15-12-9-6-3)48-57-51(54)45-42-39-36-33-30-27-23-20-17-14-11-8-5-2/h25-26,50H,4-24,27-49H2,1-3H3/b26-25- | Inchi Code 1S/C53H98O6/c1-4-7-10-13-16-19-22-25-26-29-31-34-37-40-43-46-52(55)58-49-50(59-53(56)47-44-41-38-35-32-28-24-21-18-15-12-9-6-3)48-57-51(54)45-42-39-36-33-30-27-23-20-17-14-11-8-5-2/h16,19,25-26,50H,4-15,17-18,20-24,27-49H2,1-3H3/b19-16+,26-25+ |
Still not finding the right product?
Explore all of our products under 1,3-Dipalmitoyl-2-Oleoylglycerol
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?