Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 22,60 |
Über diesen Artikel
€ 22,60
material
borosilicate glass flask, round bottom , single-neck
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F302425
capacity
25 mL, 25 mL
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description 25 mL, joint: ST/NS 24/40 | description 250 mL, joint: ST/NS 24/40 | description 2000 mL, joint: ST/NS 24/40 | description 500 mL, joint: ST/NS 24/40 |
| capacity 25 mL , 25 mL | capacity 250 mL , 250 mL | capacity 2,000 mL , 2000 mL , 2000 mL | capacity 500 mL, 500 mL |
| manufacturer/tradename Synthware F302425 | manufacturer/tradename Synthware F304250 | manufacturer/tradename Synthware F30242L | manufacturer/tradename Synthware F304500 |
| joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 |
| material single-neck , round bottom , borosilicate glass flask | material single-neck , round bottom , borosilicate glass flask | material borosilicate glass flask, single-neck , round bottom | material single-neck , borosilicate glass flask, round bottom |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ single-neck round-bottom flask
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?