Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 91,70 |
Über diesen Artikel
€ 91,70
material
borosilicate glass flask, round bottom , single-neck (high vacuum valve)
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F490010
capacity
10 mL, 10 mL
valve size
0-4 mm
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description 10 mL, valve size 0-4 mm | description 1000 mL, valve size 0-8 mm | description 250 mL, valve size 0-8 mm | description 500 mL, valve size 0-4 mm |
| manufacturer/tradename Synthware F490010 | manufacturer/tradename Synthware F49081L | manufacturer/tradename Synthware F490258 | manufacturer/tradename Synthware F490500 |
| capacity 10 mL , 10 mL | capacity 1000 mL, 1000 mL , 1,000 mL | capacity 250 mL , 250 mL | capacity 500 mL , 500 mL |
| material borosilicate glass flask, round bottom , single-neck (high vacuum valve) | material single-neck (high vacuum valve), round bottom , borosilicate glass flask | material borosilicate glass flask, single-neck (high vacuum valve), round bottom | material borosilicate glass flask, round bottom , single-neck (high vacuum valve) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| valve size 0-4 mm | valve size 0-8 mm | valve size 0-8 mm | valve size 0-4 mm |
Still not finding the right product?
Explore all of our products under Synthware™ storage flask with high vacuum valve and side arm
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?