Direkt zum Inhalt
Merck

Fortfahren mit

14-814

UbcH8 Conjugating Enzyme, 100 µg

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
12352204
NACRES:
NA.32
eCl@ss:
32160405
Biological source:
human
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


biological source

human

Quality Segment

mol wt

Mw 18 kDa

manufacturer/tradename

Upstate®

NCBI accession no.

UniProt accession no.

General description

UbcH8 is a class I ubiquitin-conjugating (E2) enzyme highly similar in structure to UbcH7. UbcH8 is a primary target gene of interferon-alpha and gamma, and recent studies have indicated that UbcH8 is the E2 enzyme for the ubiquitin-like protein ISG15. These studies suggest overlap between the ISG15 and ubiquitin conjugation pathways, and have important implications for understanding the regulation and function of ISG15 in the IFN alpha/beta response. Typical enzyme concentration to support conjugation in vitro is 100nM to 1μM depending upon conditions.
expressed in E. coli

Application

UbcH8 is a class I ubiquitin-conjugating (E2) enzyme highly similar in structure to UbcH7. UbcH8 is a primary target gene of interferon-alpha & gamma.

Physical form

Format: Purified using Ni2+ chelation chromatography with Ni2+ in limiting concentration.

Preparation Note

For long term storage store at -80°C.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial


Lagerklasse

12 - Non Combustible Liquids



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen




Questions

Reviews

No rating value

Active Filters