69418
Synonym(e):
cloning vector, expression vector, vector
Eigenschaften
100
Beschreibung
10 - Combustible liquids
WGK 3
Hier finden Sie alle aktuellen Versionen:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.
Die Dokumentenbibliothek aufrufen
This product is provided as purified plasmid DNA with a quantity of 10 µg per package.
Helpful?