Über diesen Artikel
Empirische Formel (Hill-System):
C4H4O3
CAS-Nummer:
Molekulargewicht:
100.07
UNSPSC Code:
12352108
MDL number:
vapor pressure
1.3 hPa ( 92 °C)
Quality Segment
form
scales
autoignition temp.
480 °C
potency
1510 mg/kg LD50, oral (Rat)
pH
2.7 (10 g/L in H2O)
bp
261 °C/1013 hPa
mp
116-120 °C
transition temp
flash point 157 °C
density
1.23 g/cm3 at 20 °C
bulk density
550‑600 kg/m3
storage temp.
2-30°C
InChI
1S/C4H4O3/c5-3-1-2-4(6)7-3/h1-2H2
InChI key
RINCXYDBBGOEEQ-UHFFFAOYSA-N
Analysis Note
Colour (visual): white
Appearance of substance (visual): scales
Assay (morpholine method): ≥ 98 %
Melting range
- lower value:
≥ 116 °C
- upper value: ≤ 120 °C
Identity (IR): passes test
Pd (Palladium) (ICP MS): ≤ 1 ppm
Appearance of substance (visual): scales
Assay (morpholine method): ≥ 98 %
Melting range
- lower value:
≥ 116 °C
- upper value: ≤ 120 °C
Identity (IR): passes test
Pd (Palladium) (ICP MS): ≤ 1 ppm
Still not finding the right product?
Explore all of our products under Succinic anhydride
Lagerklasse
8B - Non-combustible corrosive hazardous materials
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?