Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
eCl@ss:
36101790
UNSPSC Code:
41104105
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
stainless steel rack
Quality Segment
sterility
non-sterile
product line
NovaSeptum®
packaging
pkg of 1 unit
rack L × W × H
250 mm (9.8 in.) × 30 mm (1.2 in.) × 260-460 mm (10.2-18.1 in.)
weight
0.5 kg (1.1 lb ), (rack)
application(s)
sterile sampling
compatibility
for use with 9-port holder, TC 64 for NAC-USI, NAC-USM
General description
The NovaSeptum® Bag Rack for Single Sampling Units is a stainless steel mechanical support rack designed to hold and organize NovaSeptum® sampling units during aseptic fluid sampling operations. The rack is engineered for compatibility with the 9-port TC 64 (Tri-Clamp 64 mm) holders compatible with NAC-USI and NAC-USM (NovAseptic Connector) connectors. The rack is fully adjustable, reusable, and available for all standard holders.
Application
- The NovaSeptum® Bag Rack acts as a structured mechanical support frame that holds and organizes NovaSeptum® sampling units during aseptic fluid sampling operations.
- It is particularly suited for use in biopharmaceutical manufacturing and cleanroom environments where controlled, contamination-free fluid sampling is required.
Features and Benefits
- Fully adjustable to accommodate different process setups.
- The maximum load capacity of the rack goes up to 5 kgs.
- The structural components are manufactured from stainless steel in compliance with ASTM® 316, while the screws and locking nuts are made from A4-grade stainless steel.
- A certificate of quality is included in each package for traceability and compliance.
- Compatible with bag, bottle and conical tube sampling units
Packaging
A complete bag rack unit, including all components, is packaged unassembled in a cardboard box and assigned a part number.
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under NovaSeptum® Single-sampling Unit Bag-Rack
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.