Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
200 μL | Warenkorb auf Verfügbarkeit prüfen | € 815,00 |
Über diesen Artikel
conjugate
unconjugated
Quality Segment
antibody form
affinity purified immunoglobulin
clone
polyclonal
species reactivity
human, rat, rabbit
concentration
(Please refer to the Certificate of Analysis for the lot-specific concentration.)
technique(s)
immunohistochemistry: suitable, western blot: suitable
UniProt accession no.
target post-translational modification
unmodified
General description
Application
Immunohistochemistry on rat brain sections.
Dilutions should be made using a carrier protein such as BSA (1-3%)
Optimal working dilutions must be determined by the end user.
Biochem/physiol Actions
SPECIES REACTIVITIES: The epitope is also highly conserved between mouse and rat (17/18 residues identical) but other species have not been tested.
Physical form
Preparation Note
Other Notes
SIZE: 43518 Da [This is the MW of the unprocessed precursor].
SUBCELLULAR LOCATION: Membrane; Multi-pass membrane protein (Potential).
TISSUE SPECIFICITY: Widespread expression in adult. Strongest expression in pancreas and placenta. Lower expression in brain, lung, prostate, heart, kidney, uterus, small intestine and colon.
MISCELLANEOUS: Inhibited by external acidification. Activated by halothane and isoflurane.
SIMILARITY: Belongs to the two pore domain potassium channel (TC 1.A.1.8) family.
Disclaimer
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description - | description affinity isolated antibody, lyophilized powder | description clone S374-48, purified immunoglobulin | description Chemicon®, from rabbit |
| clone polyclonal | clone polyclonal | clone S374-48, monoclonal | clone polyclonal |
| species reactivity human, rat, rabbit | species reactivity human, rat | species reactivity mouse, rat | species reactivity rat |
| antibody form affinity purified immunoglobulin | antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form affinity purified immunoglobulin |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| technique(s) western blot: suitable, immunohistochemistry: suitable | technique(s) western blot (chemiluminescent): 1:200 | technique(s) immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable | technique(s) immunohistochemistry: suitable, western blot: suitable |
Lagerklasse
11 - Combustible Solids
Was ist los?
WGK 3
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.



