Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenconjugate
unconjugated
Quality Segment
antibody form
affinity purified immunoglobulin
clone
polyclonal
purified by
affinity chromatography
species reactivity
rabbit, human
concentration
(Please refer to lot specific datasheet.)
technique(s)
western blot: suitable
UniProt accession no.
target post-translational modification
unmodified
General description
KLH-conjugated linear peptide corresponding to the transactivation domain of human E2F-1.
~52 kDa observed
Application
Epitope: Transactivation Domain
Physical form
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Western Blotting in SW-480 cell lysate.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected E2F-1 in 10 µg of SW-480 cell lysate.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected E2F-1 in 10 µg of SW-480 cell lysate.
Other Notes
Human. Predicted to react with Rat, Bovine based on 100% sequence homology.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?