Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
mouse, human, rat
Application:
WB
Citations:
-
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenbiological source
rabbit
Quality Segment
clone
polyclonal
species reactivity
mouse, human, rat
technique(s)
western blot: suitable
General description
Proline-rich protein 14 (UniProt Q9BWN1) is encoded by the PRR14 gene in human (Gene ID 78994). Proline-rich protein 14 participates in the tethering of heterochromatin to the inner nuclear periphery by associating with the nuclear lamina and attaching to heterochromatin through its binding partner, heterochromatin protein 1 (HP1). After disassembly early in mitosis, Proline-rich protein 14 reassembles via binding to anaphase chromosomes through HP1, then via association with the nuclear lamina in telophase. PRR14 may thereby play a role in specifying HP1-bound heterochromatin for reattachment to the nuclear lamina at mitotic exit.
Immunogen
KLH-conjugated linear peptide corresponding to human Proline-rich Protein 14.
Application
Anti-Proline-rich Protein 14 Antibody is an antibody against Proline-rich Protein 14 for use in Western Blotting.
Analysis Note
Evaluated by Western Blotting in HeLa cell lysate.
Western Blotting Analysis: A 1:500 dilution of this antibody detected Proline-rich Protein 14 in 10 ug of HeLa cell lysate.
Western Blotting Analysis: A 1:500 dilution of this antibody detected Proline-rich Protein 14 in 10 ug of HeLa cell lysate.
Other Notes
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.