Direkt zum Inhalt
Merck

Fortfahren mit

ABE1795

Anti-PAXX

from rabbit, purified by affinity chromatography

Synonym(e):

Protein PAXX, Paralog of XRCC4 and XLF, XRCC4-like small protein, C9orf142

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PAXX(286257)

General description

Protein PAXX (UniProt: 9BUH6; also known as Paralog of XRCC4 and XLF, XRCC4-like small protein) is encoded by the C9orf142 (also known as PAXX, XLS) gene (Gene ID: 286257) in human. PAXX is a newer member of XRCC4 superfamily that predominantly localizes to the nucleus and is shown to accumulate at sites of DNA damage generated by laser microirradiation. It is involved in non-homologous end joining (NHEJ), a major pathway to repair double-strand breaks in DNA and may act as a scaffold required to stabilize the Ku heterodimer, composed of XRCC5/Ku80 and XRCC6/Ku70, at double-strand break sites where it promote the assembly and/or stability of the NHEJ machinery. PAXX residues 1-113 form a head domain, which is structurally related to those of XRCC4, XLF and SAS6. This is followed by a 31-amino-acid -helix, forming a coiled-coil with the other protomer to make a PAXX homodimer. Two isoforms of PAXX have been described that are produced by alternative splicing. (Ref.: Ochi, T., et al. (2015). Science 347(6218); 185-188).
~22 kDa observed; 21.64 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

KLH-conjugated liner peptide corresponding to 16 amino acids from the C-terminus region of human Protein PAXX.

Application

Anti-PAXX, Cat. No. ABE1795, is a highly specific rabbit polyclonal antibody that targets PAXX (C9orf142) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected PAXX in human prostate, human tonsil, and human cervical cancer tissues.
Research Category
Epigenetics & Nuclear Function

Biochem/physiol Actions

This rabbit polyclonal antibody detects PAXX protein. It targets an epitope within 16 amino acids from the C-terminus region.

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, pH7.4 with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HeLa cell nuclear extract.

Western Blotting Analysis: 2 µg/mL of this antibody detected PAXX in HeLa cell nuclear extract.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
SAB4502553SAB2107888SAB1410876
description

from rabbit, purified by affinity chromatography

description

affinity isolated antibody

description

affinity isolated antibody

description

purified immunoglobulin, buffered aqueous solution

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

technique(s)

western blot: suitable, immunohistochemistry: suitable (paraffin)

technique(s)

western blot: 1:500-1:1000, immunohistochemistry: 1:50-1:100, ELISA: 1:20000

technique(s)

immunoblotting: suitable

technique(s)

western blot: 1 μg/mL, immunofluorescence: suitable

Gene Information

human ... PAXX(286257)

Gene Information

human ... PXN(5829)

Gene Information

human ... PAX3(5077)

Gene Information

human ... PAX2(5076)

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

Probieren Sie unser Produkt-Auswahlhilfe, um Ihre Auswahl einzugrenzen


Lagerklasse

12 - Non Combustible Liquids

Was ist los?

WGK 1

Flammpunkt (°F)

does not flash

Flammpunkt (°C)

does not flash



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen




Questions

Reviews

No rating value

Active Filters