Direkt zum Inhalt
Merck

Fortfahren mit

ABE492

Anti-Histone lysine demethylase NO66 Antibody

from rabbit, purified by affinity chromatography

Synonym(e):

Bifunctional lysine-specific demethylase and histidyl-hydroxylase NO66, 60S ribosomal protein L8 histidine hydroxylase, Histone lysine demethylase NO66, Myc-associated protein with JmjC domain, Nucleolar protein 66, hsNO66, Ribosomal oxygenase NO66, ROX,

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis
100 μg
Warenkorb auf Verfügbarkeit prüfen
€ 339,00

Über diesen Artikel

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-

€ 339,00


Warenkorb auf Verfügbarkeit prüfen

​
Sonderanfertigung anfragen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RIOX1(79697)

General description

NO66 is a lysine-specific demethylase that removes methyl groups from histone H3 at lysine 4 and lysine 36. This enzyme has been shown to more readily demethylate trimethylated H3 ′Lys-4′ (H3K4me3) and monomethylated H3 Lys4 (H3K4me1) consistent with its role in modulating the histone code. This protein is a highly conserved, constitutive nucleolar component that shows a dual localization pattern with strong enrichment in the granular part of nucleoli and in distinct nucleoplasmic entities.
~66 kDa observed

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to human Histone lysine demethylase NO66 near the N-terminus.

Application

Detect NO66 using this rabbit polyclonal antibody, Anti-Histone lysine demethylase NO66 Antibody validated for use in western blotting.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Biochem/physiol Actions

This antibody recognizes Histone lysine demethylase NO66 near the N-terminus.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 cell lysate
Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Histone lysine demethylase NO66 in 10 µg of A431 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
H7403ABE498SAB4301885
description

from rabbit, purified by affinity chromatography

description

affinity isolated antibody, buffered aqueous solution

description

from rabbit, purified by affinity chromatography

description

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

UniProt accession no.

Q9H6W3

UniProt accession no.

-

UniProt accession no.

Q6ZMT4

UniProt accession no.

-

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

Probieren Sie unser Produkt-Auswahlhilfe, um Ihre Auswahl einzugrenzen


Lagerklasse

12 - Non Combustible Liquids

WGK

WGK 1



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen




Questions

Reviews

No rating value

Active Filters