Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
biological source
rabbit
Quality Segment
antibody form
purified antibody
antibody product type
primary antibodies
clone
RM155, monoclonal
species reactivity
human
technique(s)
multiplexing: suitable, western blot: suitable
isotype
IgG
UniProt accession no.
shipped in
wet ice
target post-translational modification
trimethylation (Lys36)
Gene Information
human ... H3F3B(3021)
General description
Immunogen
Application
Biochem/physiol Actions
Physical form
Analysis Note
Western Blotting Analysis (WB): 2 μg/mL of this antibody detected trimethyl Histone H3 (Lys36) in Recombinant HeLa protein and HeLa acid extract.
Other Notes
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description clone RM155, from rabbit | description clone RM140, from rabbit | description recombinant, expressed in HEK 293 cells, clone RM155, purified immunoglobulin | description from rabbit, purified by affinity chromatography |
| species reactivity human | species reactivity human | species reactivity vertebrates | species reactivity human, mouse |
| antibody form purified antibody | antibody form - | antibody form purified immunoglobulin | antibody form affinity isolated antibody |
| biological source rabbit | biological source rabbit | biological source - | biological source rabbit |
| Gene Information human ... H3F3B(3021) | Gene Information human ... H3F3B(3021) | Gene Information human ... H3C1(8350) | Gene Information human ... HIST1H3F(8968) |
| clone RM155, monoclonal | clone RM140, monoclonal | clone RM155, monoclonal, recombinant monoclonal | clone polyclonal |
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level 100 |
Still not finding the right product?
Probieren Sie unser Produkt-Auswahlhilfe, um Ihre Auswahl einzugrenzen
Lagerklasse
12 - Non Combustible Liquids
Was ist los?
WGK 2
Flammpunkt (°F)
does not flash
Flammpunkt (°C)
does not flash
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.



