Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
eCl@ss:
32160702
UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
monoclonal
Application:
—
Citations:
-
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
monoclonal
mol wt
(~75 kDa observed; 75.17 kDa calculated. Uncharacterized bands may be observed in some lysate(s).)
purified by
using protein G
species reactivity
mouse
isotype
IgG2aκ
Protein ID accession no.
UniProt accession no.
target post-translational modification
unmodified
Application
Anti-Lap2alpha, clone 8C10-1H11, Cat. No. MABF1997, is a mouse monoclonal antibody that detects Lamina-associated polypeptide 2, isoforms alpha, and has been tested for use in Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), and Western Blotting.
Immunofluorescence Immunoprecipitation Chromatin Immunoprecipitation (ChIP) Western Blotting
Western Blotting Analysis: A representative lot detected Lap2alpha in Western Blotting applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73).
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected Lap2alpha in Chromatin Immunoprecipitation applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Lap2alpha in Immunoprecipitation applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73).
Immunofluorescence Analysis: A representative lot detected Lap2alpha in Immunofluorescence applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73).
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected Lap2alpha in Chromatin Immunoprecipitation applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Lap2alpha in Immunoprecipitation applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73).
Immunofluorescence Analysis: A representative lot detected Lap2alpha in Immunofluorescence applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73).
Biochem/physiol Actions
Clone 8C10-1H11 specifically detects Lamina-associated polypeptide 2 alpha (LAP2alpha) in murine species. It targets an epitope in the internal region.
Features and Benefits
His-tagged recombinant fragment corresponding to 507 amino acids from the C-terminal region.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Other Notes
Evaluated by Western Blotting in Knockout (KO) vs. wild-type (WT) mouse fibroblast cell lysates.
Western Blotting Analysis: 1 µg/mL of this antibody detected Lamina-associated polypeptide 2, isoforms alpha (LAP2alpha) in wild-type, but not in Knockout (KO) mouse fibroblast cell lysates.
Western Blotting Analysis: 1 µg/mL of this antibody detected Lamina-associated polypeptide 2, isoforms alpha (LAP2alpha) in wild-type, but not in Knockout (KO) mouse fibroblast cell lysates.
Lamina-associated polypeptide 2, isoforms alpha (UniProt: Q61033; also known as Thymopoietin isoform alpha, TP alpha) is encoded by the Tmpo (also known as Lap2) gene (Gene ID: 21917) in murine species. LAP2alpha is a non-membrane bound, oligomeric nucleoplasmic protein that associates directly or indirectly with lamins at specific cell-cycle stages. It is expressed diffusely throughout the nucleus and is involved in the structural organization of the nucleus and in the post-mitotic nuclear assembly. It is shown to interact with A-type lamins to control gene expression, transcription, and chromatin organization. The N-terminal part of LAP2alpha contains two structurally independent, non-interacting domains: LEM (LAPEmerin-MAN1)-like domain that binds DNA and a LEM domain that interacts with BANF1. Its C-terminal domain forms a four-stranded coiled coil. Lamin A/C and LAP2alpha complexes predominantly co-occur in euchromatic genomic regions and are shown to be highly mobile. They can bind chromatin in a dynamic manner and are involved in establishing a chromatin environment that is permissive for epigenetic regulation. Loss of LAP2alpha has been linked to increased proliferation of tissue progenitor cells in epidermis, colon, and the hematopoietic system and delayed skeletal muscle differentiation. (Res.: Gesson, K et al., (2016). Genome Res. 26(4);462-473).
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Probieren Sie unser Produkt-Auswahlhilfe, um Ihre Auswahl einzugrenzen
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.