Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
7H4.1, monoclonal
Application:
IHC, WB
Citations:
-
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenconjugate
unconjugated
Quality Segment
antibody form
purified antibody
clone
7H4.1, monoclonal
purified by
using protein G
species reactivity
mouse, human
concentration
(Please refer to lot specific datasheet.)
technique(s)
immunohistochemistry: suitable, western blot: suitable
isotype
IgG1κ
UniProt accession no.
target post-translational modification
unmodified
General description
GST-tagged recombinant protein corresponding to human PER2.
~136 kDa observed
Application
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected PER2 in human pancreas and human kidney tissue.
Physical form
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Western Blotting in HepG2 nuclear extract.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected PER2 in 200 µg of HepG2 nuclear extract.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected PER2 in 200 µg of HepG2 nuclear extract.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Lagerklasse
12 - Non Combustible Liquids
Was ist los?
WGK 1
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.