Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
eCl@ss:
32160702
UNSPSC Code:
12352203
Species reactivity:
mouse
Technique(s):
ELISA: suitable
Application:
ELISA
Citations:
-
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenantibody form
purified antibody
Quality Segment
assay
95% (SDS-PAGE)
purified by
using Protein A
species reactivity
mouse
concentration
25 mg/mL
technique(s)
ELISA: suitable
isotype
IgG
Physical form
Non-sterile liquid in 0.1M Phosphate buffer, pH 7.7 filtered through 0.2 µm filter.
Preparation Note
Stable for 1 year at from date of receipt.
Analysis Note
ELISA: A previous lot of this antibody was used in ELISA.
Disclaimer
This product is intended for use by IVD manufacturers.
Contact customer service for assistance: oem.reagents@emdmillipore.com
Contact customer service for assistance: oem.reagents@emdmillipore.com
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?