Direkt zum Inhalt
Merck

Fortfahren mit

SCC0831

ReNcell® Human Neural Progenitor Cell Line (ReNSC1)

Human neural stem cell (NSC) line that differentiates into neuronal and glial subtypes. Normal, Cortex, Age: Fetal, Male, Mutation: None

Synonym(e):

ReNcell® Human Neural Stem Cell Line

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
41106514
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


Quality Segment

biological source

human

packaging

vial of ≥1X10⁶ cells/vial vial

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell culture | mammalian: suitable

storage temp.

-140 to -196°C

General description

Understanding human neurodevelopment and the underlying biology remains a complex issue. Stem cell biology improvements have introduced new tools and more useful model systems. Human neural stem cells (hNSCs) and neural progenitor cells (NPCs) have provided a robust system in which to investigate the complex neurobiology questions in modern science. NPCs have the capability to differentiate into different neural cell types such as astrocytes and oligodendrocytes. They are useful in screening new, potentially therapeutic drugs in a highly cost-efficient and high-throughput manner due to the reliable and robust source of cells. NPCs can also be easily manipulated, making them an ideal platform to use.1 The ReNcell® Human Neural Progenitor group of cell lines are conditionally immortalized human neural progenitor cells which appropriately retain characteristics of their original tissue sites. Furthermore, the cells are genetically modified with a chemical switch (conditional immortalization) to give a greater control of proliferation and differentiation. While these cells are grown in their progenitor state, they can be terminally differentiated into neurons with distinct characteristics reflective of their original brain regions. This makes the ReNcell® hNPCs a strong source of distinct neural cells in which to further investigate neurobiology. Each line was conditionally immortalized using retroviral integration of a single copy of the c-mycERTAM construct.1 This allows the cells to retain proliferative properties when grown with the necessary growth factors and 4-hydroxytamoxifen (4-OHT). With changes in media formulation, the cells start to differentiate into mature neurons characteristic of their original tissue region. The ReNSC1 (SCC0831) cell line is originally sourced from human cortex. The ReNcell® hNPC lines have a high number of potential applications in which they can be utilized such as disease modelling, drug discovery, neural cytotoxicity testing, and target validation. 

Application

  • Specifications
  • • Viability: > 70% viable upon thaw
  • • Cell Number: ≥1X106 cells/vial
  • • Passage Expansion: > Expand for 5 passages
  • • Species Verification: Species has been verified and confirmed to be negative for inter-species contamination by a Contamination Clear panel by Charles River Animal Diagnostic Services.
  • • Viral or Infectious Disease Testing: Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • • Microbial Contamination: None detected.
  • • Mycoplasma Contamination: Negative
  • • STR Profile: Pass
  • • Cell Morphology: See Images
  • • Characterization: Immunocytochemistry

Preparation Note

These cells should be stored in liquid nitrogen.

Analysis Note

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Other Notes

1.Anderson GW, Deans M, Taylor RDT, Raval P, Chen D, Lowder H, Srishti Murkerji, Andreae LC, Williams BP, Srivastava DP. 2015. Characterisation of neurons derived from a cortical human neural stem cell line CTX0E16. Stem Cell Research & Therapy. 6(1). doi:https://doi.org/10.1186/s13287-015-0136-8. [accessed 2024 Sep 26]. https://stemcellres.biomedcentral.com/articles/10.1186/s13287-015-0136-8.2.Jeffries AR, Perfect LW, Ledderose J, Schalkwyk LC, Bray NJ, Mill J, Price J. 2012. Stochastic Choice of Allelic Expression in Human Neural Stem Cells. STEM CELLS. 30(9):1938–1947. doi:https://doi.org/10.1002/stem.1155.3.Anacker C, Zunszain PA, Cattaneo A, Carvalho LA, Garabedian MJ, Thuret S, Price J, Pariante CM. 2011. Antidepressants increase human hippocampal neurogenesis by activating the glucocorticoid receptor. Molecular Psychiatry. 16(7):738–750. doi:https://doi.org/10.1038/mp.2011.26.4.Miljan EA, Hines SJ, Pande P, Corteling RL, Hicks C, Zbarsky V, Umachandran M, Sowinski P, Richardson S, Tang E, et al. 2009. Implantation of c-mycERTAMImmortalized Human Mesencephalic-Derived Clonal Cell Lines Ameliorates Behavior Dysfunction in a Rat Model of Parkinson′s Disease. Stem Cells and Development. 18(2):307–320. doi:https://doi.org/10.1089/scd.2008.0078.5.Amemori T, Romanyuk N, Jendelova P, Herynek V, Turnovcova K, Prochazka P, Kapcalova M, Cocks G, Price J, Sykova E. 2013. Human conditionally immortalized neural stem cells improve locomotor function after spinal cord injury in the rat. Stem Cell Research & Therapy. 4(3):68. doi:https://doi.org/10.1186/scrt219.

Legal Information

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/restrictedcelluse. This product may not be: re-engineered or copied; used to make derivatives, modifications or functional equivalents; used to obtain patents or other IP claiming use of the product; used to develop, test, or manufacture a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact licensing@milliporesigma.com

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Lagerklasse

10 - Combustible liquids

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable


Zulassungslistungen

Zulassungslistungen werden hauptsächlich für chemische Produkte erstellt. Für nicht-chemische Produkte können hier nur begrenzte Angaben gemacht werden. Kein Eintrag bedeutet, dass keine der Komponenten gelistet ist. Es liegt in der Verantwortung des Benutzers, die sichere und legale Verwendung des Produkts zu gewährleisten.

10043-52-4 + 7786-30-3

CAS No.


Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen