Direkt zum Inhalt
Merck

Fortfahren mit

SCC655

NM11 Low-Metastatic Murine Cell Line

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
41106514
NACRES:
NA.81
Biological source:
mouse
Manufacturer/tradename:
Millipore
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


biological source

mouse

Quality Segment

packaging

vial of ≥1X10⁶ vial (viable cells per vial)

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

General description

Tumor metastasis is one of the most life-threatening events in cancer and often leads to inevitable cancer growth that cannot be effectively treated. Treatment options after metastasis often involve slowing tumor growth or relieving harsh symptoms. The process of metastasis, known as the metastatic cascade, involves a series of events in which cancer cells break away from the primary tumor and begin growing in other tissues. This process proceeds by cancer cells moving through blood vessels or lymph nodes and spreading via the circulatory system to eventually colonize other more distant tissues. The importance of understanding metastasis remains continually relevant and requires a variety of models for understanding the underlying mechanisms. LuM1 (SCC654) and NM11 (SCC655) were developed together to provide two similar cell models with differing metastatic potentials. NM11 was selected as low-metastatic cell line and is derived from murine colon adenocarcinoma while LuM1 was developed through repeated tumor transplantation until metastatic potential of the cells had changed visibly through lung tumor nodule formation.

The LuM1 and NM11 cell lines have been utilized for metastasis, migration, and invasion assays. LuM1 and NM11 have been useful in understanding Matrix MetalloProteinase (MMP) protein expression and their relation to metastatic potential and tumor progression.

Source: Derived from mouse colon adenocarcinoma that was transplanted into a BALB/c mouse. Resulting tumor was repeatedly transplanted into new mice until metastatic potential of the tumors were visible through lung nodule formation.

References

Sakata K, Kozaki K, Iida K, Tanaka R, Yamagata S, Utsumi KR, Saga S, Shimizu S, Matsuyama M. 1996. Establishment and characterization of high- and low-lung-metastatic cell lines derived from murine colon adenocarcinoma 26 tumor line. Japanese journal of cancer research : Gann. 87(1):78–85.

Taha EA, Chiharu Sogawa, Yuka Okusha, Kawai H, May Wathone Oo, Abdellatif Elseoudi, Lu Y, Hitoshi Nagatsuka, Kubota S, Ayano Satoh, et al. 2020. Knockout of MMP3 Weakens Solid Tumor Organoids and Cancer Extracellular Vesicles. Cancers. 12(5):1260–1260.

Application

  • Cells are verified to be of mouse origin and negative for human, rat, Chinese hamster, Golden Syrian hamster, and non-human primate interspecies contamination, as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
  • Cells tested negative for infectious diseases against a Mouse Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells tested negative for mycoplasma.

Features and Benefits

NM11 cells have been utilized for metastasis, migration, and invasion assays. NM11 cells have been useful in understanding Matrix MetalloProteinase (MMP) protein expression and their relation to metastatic potential and tumor progression.

Preparation Note

The cells should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
M9269M7769M7894
description

-

description

clone MOPC 21, purified immunoglobulin, buffered aqueous solution

description

clone UPC 10, ascites fluid, lyophilized powder

description

clone MOPC 21, ascites fluid, lyophilized powder

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

technique(s)

-

storage temp.

−196°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

shipped in

liquid nitrogen

shipped in

dry ice

shipped in

-

shipped in

-

manufacturer/tradename

Millipore

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

Quality Level

100

Quality Level

300

Quality Level

300

Quality Level

200


Lagerklasse

12 - Non Combustible Liquids

WGK

WGK 2

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen