Direkt zum Inhalt
Merck

Fortfahren mit

SCC724

3dGRO® Colorectal Organoids (CSC.449)

Adenocarcinoma, Colon, Age: 40, Male, Mutation: Not Available

Synonym(e):

Colon Organoid, Colon Tumor Organoid, Colorectal Cancer Organoid, Human CRC Organoids

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
41106514
NACRES:
NA.51
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


Quality Segment

biological source

human colon

packaging

vial of ≥1500 organoids vial (viable cells per vial)

manufacturer/tradename

Millipore

gender

male

technique(s)

cell culture: suitable

application(s)

3D cell culture

control

colon

relevant disease(s)

adenocardinoma

shipped in

liquid nitrogen

storage temp.

−196°C

General description

Colorectal cancer accounts for roughly 10% of all cancer cases worldwide with more than half of all patients with CRC developing metastatic disease leading to death. Recently, tissue derived organoids have emerged as a more predictive 3 dimensional cell culture model of disease. 3D organoid cultures conserve the original genetic and phenotypic characteristics of the primary tissue allowing for their application in many research fields included drug development, personalized medicine and potential therapeutics. In vitro cultured tumor organoids have also been shown to predict patient response to chemotherapeutics. PDOs derived from colorectal cancer (CRC organoids) have been used for cell modeling and to investigate the function of cancer related driver gene mutations including APC, TP53, KRAS, BRAF, PIK3CA etc.

We are now offering a comprehensive biobank of highly characterized tissue-derived human gastrointestinal organoids from normal and diseased patients. The intestinal organoid biobank contains over 50 highly characterized intestinal organoids from both normal and diseased patients derived from multiple regions of the digestive systems including small intestine (duodenum, ileum), stomach, rectum and colon.

<bold>Cell Line Characteristics</bold>
  • Sex: Male
  • Age: 40 Years
  • Organ: Colon (Primary Tumor)
  • Disease: Adenocarcinoma. Matched pair with (CSC.449N) (SCC727)

Features and Benefits

Cryopreserved patient derived colorectal cancer organoids (PDOs)

Preparation Note

Store in liquid nitrogen. The organoids can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Legal Information

3dGRO® is a trademark of Merck KGaA, Darmstadt, Germany.
3dGRO® organoids were derived utilizing HUB Organoid Technology.

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/pdouse. This product may not be re-engineered or copied; used to make derivatives or modifications; used to develop, test, or manufacture a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact organoids@milliporesigma.com.
3dGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
SCC727SCC735SCC741
description

Adenocarcinoma, Colon, Age: 40, Male, Mutation: Not Available

description

Normal Adjacent (CRC), Colon, Age: 40, Male, Mutation: None

description

Adenocarcinoma, metastatic (liver), Colon, Age: 61, Male, Mutation: Not Available

description

Adenocarcinoma, Colon, Age: 43, Male, Mutation: Not Available

biological source

human colon

biological source

human colon

biological source

human colon

biological source

human colon

relevant disease(s)

adenocardinoma

relevant disease(s)

(normal adjacent CRC)

relevant disease(s)

adenocardinoma (metastatic liver)

relevant disease(s)

adenocardinoma

application(s)

3D cell culture

application(s)

3D cell culture

application(s)

3D cell culture

application(s)

3D cell culture

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

packaging

vial of ≥1500 organoids vial (viable cells per vial)

packaging

vial of ≥1500 organoids vial

packaging

vial of ≥1500 organoids vial

packaging

vial of ≥1500 organoids vials


Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen