Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
biological source
human
Quality Segment
packaging
vial of 50 mL
manufacturer/tradename
Millipore
technique(s)
cell culture | stem cell: suitable
shipped in
dry ice
storage temp.
−20°C
General description
Application
- Appearance (Color): Clear/No Particlulates (Red Liquid)
- Osmolality: 350-375 mOsm
- pH: 7.0 – 7.4
- Sterility Tested: No Growth/Pass
- Endotoxin: 2 EU/mL
- Mycoplasma: Negative
- Functional Assay: Thaw and culture of human iPSC derived liver organoids for 2 passages.
Features and Benefits
Preparation Note
Other Notes
Legal Information
Disclaimer
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description - | description Normal, Dermal Fibroblast, Age: Neonatal, Male, Mutation: None | description Normal, Liver, Age: 63, Female, Mutation: None | description Normal, Liver, Age: 57, Female, Mutation: None |
| technique(s) cell culture | stem cell: suitable | technique(s) cell culture: suitable | technique(s) cell culture: suitable | technique(s) cell culture: suitable |
| storage temp. −20°C | storage temp. −196°C | storage temp. −196°C | storage temp. −196°C |
| shipped in dry ice | shipped in liquid nitrogen | shipped in liquid nitrogen | shipped in liquid nitrogen |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| packaging vial of 50 mL | packaging vial of ≥1500 vial (viable cells per vial) | packaging vial of 1 vial >=1500 viable cells per vial | packaging vial of 1 vial >=1500 viable cells per vial |
| biological source human | biological source human (dermal fibroblast) | biological source human liver | biological source human liver |
Lagerklasse
12 - Non Combustible Liquids
Was ist los?
WGK 3
Flammpunkt (°F)
No data available
Flammpunkt (°C)
No data available
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.



