Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
UNSPSC Code:
41106200
NACRES:
NA.42
eCl@ss:
32160703
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenProduktname
EPIGRO® Human Prostate Epithelia Basal Medium,
form
liquid
Quality Segment
storage temp.
2-8°C
Application
To form a complete kit both components are required: Basal Medium (Catalog No. SCMP-BM) and Supplement Mix (Catalog No. SCMP001-S).
EpiGRO® Prostate Epithelia Complete Media Kit is optimized for the culture of human prostate epithelial cells. It supports the growth of these cells over a period of at least 15 population doublings. This medium does not contain any antimicrobials or phenol red; components that can cause cell stress and “masking effects” that may influence experimental results. It is formulated for use in an incubator with an atmosphere of 5% CO⒉
EpiGRO® Prostate Epithelia Complete Media Kit is optimized for the culture of human prostate epithelial cells. It supports the growth of these cells over a period of at least 15 population doublings. This medium does not contain any antimicrobials or phenol red; components that can cause cell stress and “masking effects” that may influence experimental results. It is formulated for use in an incubator with an atmosphere of 5% CO⒉
Preparation Note
Basal Medium should be stored at 2-8ºC in the dark. Do not freeze. Supplement Mix should be stored at -20ºC. Supplemented Medium is stable for up to 4 weeks at 2-8ºC. Do not freeze the supplemented medium. For use, aliquot and pre-warm the needed amount of medium, keeping the remaining medium refrigerated at 2-8ºC.
Analysis Note
Each lot is checked for growth promoting activity, adherence rate, and typical morphology. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Other Notes
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Legal Information
EPIGRO is a registered trademark of Merck KGaA, Darmstadt, Germany
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description - | description - | description - | description - |
| form liquid | form frozen liquid | form liquid | form frozen liquid |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C | storage temp. −20°C |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
Lagerklasse
12 - Non Combustible Liquids
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Zulassungslistungen
Zulassungslistungen werden hauptsächlich für chemische Produkte erstellt. Für nicht-chemische Produkte können hier nur begrenzte Angaben gemacht werden. Kein Eintrag bedeutet, dass keine der Komponenten gelistet ist. Es liegt in der Verantwortung des Benutzers, die sichere und legale Verwendung des Produkts zu gewährleisten.
10035-04-8 + 7783-00-8 + 67-48-1
CAS No.
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.



