Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
UNSPSC Code:
41106514
NACRES:
NA.76
eCl@ss:
32011203
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenQuality Segment
description
Cell line type: Neural Stem Cells, Neural Stem Cells
technique(s)
cell culture | stem cell: suitable, immunocytochemistry: suitable
General description
The Mouse Spinal Cord Neural Stem Cell Expansion Kit provides a multi-component system for the culture and analysis of neural stem cells and their differentiated progenies. This system includes primary neural stem cells isolated from the spinal cord of embryonic day 15-18 (E15-E18) C57/BL6 mice, neural stem cell expansion medium, and a panel of antibodies for immunocytochemical staining of neural stem/progenitor cells (Nestin and Sox2) and differentiated neural phenotypes (Map2ab for neurons, GFAP for astrocytes, and O1 for oligodendrocytes).
Other Notes
Cryopreserved Mouse Spinal Cord Neural Stem Cells (Cat.# SCR031) Mouse Neural Stem Cell Expansion Medium (Cat.# SCM008) Neural Stem Cell Marker Characterization Kit (Cat.# SCR019)
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Lagerklasse
10 - Combustible liquids
Was ist los?
WGK 2
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?