Direkt zum Inhalt
Merck

Fortfahren mit

UPMPCFDNA

Ultra pure MagPrep® cfDNA Isolation Kit

No proteinase K or heating step needed for Magprep® Nucleic Acid Isolation,Nuclease free (NICKase, RNase, DNase), DNA-free, High-throughput Compatible and Manual

Nachhaltigkeitsmerkmale

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
41105500
NACRES:
NA.51
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


Quality Segment

usage

sufficient for 100 purifications

sustainability

Sustainability features

concentration

45-55 mg/mL (Magnetic particle)

test parameters

total test time: 5 min

suitability

suitable for nucleic acid purification

greener alternative category

storage temp.

2-30°C

General description

Click here to view its DfS scorecard.

The Ultra pure MagPrep® cfDNA isolation kit is designed for highly efficient cfDNA nucleic acid isolation from most biological fluids in both manual or high-throughput automated platforms. The kit contains magnetic silica particles and uniquely formulated buffers optimized for rapid easy handling and maximal yield. Nucleic acid isolation is achieved without the need for Proteinase K or sample heating, and all items are shipped and stored at ambient temperature. All kit components are quality tested to be free of contaminant nucleases and DNA, reducing the likelihood of false positives or negatives in downstream assays. The purified nucleic acid eluate is free of PCR inhibitors such as chaotropic salts or alcohols, allowing for effective amplification in various downstream applications such as RT-qPCR, ddPCR, NGS, and cDNA generation. For research or further manufacturing use only. Not intended for direct use in humans or animals. Non-TSCA use only in US.

Features and Benefits

  • Nucleic Acid Isolation
  • Manual or high-throughput automated isolations
  • RT-qPCR
  • Digital PCR
  • Biological samples can be used
  • Room temperature stable reagents
  • DNA free
  • DNase free
  • RNase free
  • NICKase free
  • simple and fast protocols

Legal Information

MAGPREP is a registered trademark of Merck KGaA, Darmstadt, Germany

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
UPMPCFDBBUPMPVRNAUPMPVRBB
description

No proteinase K or heating step needed for Magprep® Nucleic Acid Isolation,Nuclease free (NICKase, RNase, DNase), DNA-free, High-throughput Compatible and Manual

description

No proteinase K or heating step needed for Magprep® Nucleic Acid Isolation,Nuclease free (NICKase, RNase, DNase), DNA-free, High-throughput Compatible and Manual

description

No proteinase K or heating step needed for Magprep® Nucleic Acid Isolation,Nuclease free (NICKase, RNase, DNase), DNA-free, High-throughput Compatible and Manual

description

-

usage

sufficient for 100 purifications

usage

1000 purifications

usage

sufficient for 100 purifications

usage

1000 purifications

test parameters

total test time: 5 min

test parameters

total test time: 5 min

test parameters

total test time: 5 min

test parameters

total test time: 5 min

sustainability

Greener Alternative Product, Greener Alternative Product

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

suitability

suitable for nucleic acid purification

suitability

suitable for nucleic acid purification

suitability

suitable for nucleic acid purification

suitability

suitable for nucleic acid purification

concentration

45-55 mg/mL (Magnetic particle)

concentration

45-55 mg/mL (Magnetic particle)

concentration

45-55 mg/mL (Magnetic particle)

concentration

45-55 mg/mL (Magnetic particle)


Nur Kit-Komponenten

Produkt-Nr.
Beschreibung

  • UPMPPART-2ML-KC Ultra pure MagPrep Magnetic Particles 1

  • UPMPCFDBB-50ML-KC Ultra pure MagPrep cfDNA Binding Buffer 1

  • UPMPWB1-100ML-KC Ultra pure MagPrep Wash Buffer 1 1

  • UPMPWB2-100ML-KC Ultra pure MagPrep Wash Buffer 2 1

  • UPMPEB-10ML-KC Ultra pure MagPrep Elution Buffer 1

Signalwort

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Zielorgane

Thyroid

Lagerklasse

3 - Flammable liquids

Was ist los?

WGK 3



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen