Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
UNSPSC Code:
12352200
NACRES:
NA.21
eCl@ss:
32160702
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltengrade
analytical standard
format
neat
storage temp.
−20°C
General description
Source:
Hazelnut Flour Formulation:
Preservative and carrier-free in sterile, endotoxin-free Phosphate-buffered saline, pH 7.4. Sterile filtered.
Notes:
(1) A LoToxTM product: Endotoxin ≤ 0.03 EU/µg.
(2) Relative abundance results are based on label-free quantification of precursor peptide ion intensity (peak area, incl. unique + razor peptides) using Proteome Discoverer 2.2 algorithms.
(3) Calculations of relative abundance can be variable and depend on multiple factors such as sample complexity, instrument resolution and analysis software.
Hazelnut Flour Formulation:
Preservative and carrier-free in sterile, endotoxin-free Phosphate-buffered saline, pH 7.4. Sterile filtered.
Notes:
(1) A LoToxTM product: Endotoxin ≤ 0.03 EU/µg.
(2) Relative abundance results are based on label-free quantification of precursor peptide ion intensity (peak area, incl. unique + razor peptides) using Proteome Discoverer 2.2 algorithms.
(3) Calculations of relative abundance can be variable and depend on multiple factors such as sample complexity, instrument resolution and analysis software.
Disclaimer
For research and commercial use in vitro: not for human in vivo or therapeutic use.
Lagerklasse
10 - Combustible liquids
WGK
WGK 1
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?