Fortfahren mit
Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
product line
ISOM8™
Quality Segment
assay
(HPLC), ≥98%
form
solid
mol wt
468.69
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction, reaction type: Cross Couplings, reaction type: Heck Reaction, reaction type: Hiyama Coupling, reaction type: Negishi Coupling, reaction type: Sonogashira Coupling, reaction type: Stille Coupling, reaction type: Suzuki-Miyaura Coupling, reagent type: ligand
mp
121-131 °C
application(s)
diagnostic assay manufacturing
functional group
phosphine
storage temp.
room temp
SMILES string
CC1=C(C2=C(C(C)C)C=C(C(C)C)C=C2C(C)C)C(P(C(C)(C)C)C(C)(C)C)=C(OC)C=C1
InChI
1S/C31H49OP/c1-19(2)23-17-24(20(3)4)28(25(18-23)21(5)6)27-22(7)15-16-26(32-14)29(27)33(30(8,9)10)31(11,12)13/h15-21H,1-14H3
InChI key
CVLLAKCGAFNZHJ-UHFFFAOYSA-N
General description
Our portfolio of ISOM8 products and chemistries is ever evolving to support our valued Sigma-Aldrich® customers′ requirements. Please navigate to our documents section to look at dossiers and other quality documentation for this product, provided with support of our Elevate Program.
We are committed to bringing you Greener Alternative Products, which adhere to one of thefour categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Atom Economy” and “Design for Energy Efficiency”. Click here to view its DOZN scorecard.
Application
Recent Studies and Innovations:
Mild and general C–O coupling of primary alcohols: Zhang et al. reported two complementary RockPhos-based catalyst systems providing mild and general conditions for the Pd-catalyzed C–O cross-coupling of primary alcohols with aryl and heteroaryl halides. The RockPhos Pd G3 precatalyst was specifically demonstrated for activated substrates, affording aryl ethers in high yields with excellent functional-group tolerance. (Zhang et al., 2018)
Improved process for secondary alcohol C–O cross-coupling: Zhang, Ruiz-Castillo, Schuppe, and Buchwald described an improved protocol for the Pd-catalyzed C–O cross-coupling of secondary alcohols with (hetero)aryl chlorides. The use of RockPhos as the ligand was key to achieving efficient coupling with only a 20% molar excess of alcohol, a significant advance over prior methods requiring stoichiometric excess and elevated temperatures. (Zhang et al., 2020)
RockPhos is a sterically demanding dialkylbiaryl monophosphine ligand developed by the Buchwald group, designed to enable challenging palladium-catalyzed carbon–oxygen (C–O) cross-coupling reactions, as well as C–N and other bond-forming reactions with sterically demanding substrates.
Notable Characteristics & Key Uses
- High Steric Bulk and Electron Richness: RockPhos combines adi-tert-butylphosphino donor with a bulky tri-isopropyl/methoxy biarylbackbone, delivering a large cone angle that accelerates both oxidative addition of hindered aryl electrophiles and reductive elimination from palladium intermediates.
- Selectivity for C–O Bond Formation: RockPhos is particularlyoptimized for the intermolecular C–O coupling of aryl halides with primary andsecondary aliphatic alcohols, a transformation that is challenging with mostother phosphine ligands due to competing β-hydride elimination.
- Broad Substrate Scope: The ligand enables coupling across awide range of aryl and heteroaryl halides (chlorides, bromides, and iodides)with structurally diverse alcohol nucleophiles, including those bearingsensitive functional groups and fluoroalkyl substituents.
- Low Palladium Loading: High catalyst turnover allowsreactions to proceed at palladium loadings as low as 0.5 mol%, reducing metalwaste and simplifying downstream purification, which is critical forpharmaceutical applications.
- Aryl ether synthesis: RockPhos supports palladium inthe formation of C–O bonds between aryl halides and primary or secondaryaliphatic alcohols, providing efficient access to diaryl and alkyl aryl ethersthat are prevalent motifs in pharmaceuticals and agrochemicals.
- Coupling of primary alcohols under mild conditions: Using the RockPhos Pd G3 precatalyst, primary aliphatic alcohols coupleefficiently with diverse aryl and heteroaryl halides in good to excellentyields at moderate temperatures, making the method amenable to complex,multifunctional substrates.
- Improved C–O cross-coupling of secondary alcohols: RockPhos enables the challenging palladium-catalyzed C–O coupling of secondaryalcohols with (hetero)aryl chlorides using near-equimolar amounts of thealcohol partner, an advancement over earlier systems that required large excesses and higher temperatures.
- Synthesis of fluorinated alkyl aryl ethers: RockPhos-basedcatalysis provides access to fluorinated ether motifs via C–O cross-coupling offluoroalkyl alcohols with aryl halides, delivering building blocks of highrelevance for medicinal chemistry and drug discovery.
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description ≥98%, ISOM8™ | description 97% | description - | description - |
| reaction suitability reaction type: Suzuki-Miyaura Coupling, reaction type: Cross Couplings, reaction type: Heck Reaction, reaction type: Buchwald-Hartwig Cross Coupling Reaction, reaction type: Hiyama Coupling, reaction type: Negishi Coupling, reagent type: ligand, reaction type: Sonogashira Coupling, reaction type: Stille Coupling | reaction suitability reaction type: Negishi Coupling, reagent type: ligand | reaction suitability reaction type: Stille Coupling, reagent type: catalyst | reaction suitability reaction type: Buchwald-Hartwig Cross Coupling Reaction, reagent type: catalyst |
| functional group phosphine | functional group phosphine | functional group phosphine | functional group phosphine, (Palladium) |
| assay (HPLC), ≥98% | assay 97% | assay - | assay - |
| Quality Level 300 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| form solid | form solid | form solid | form powder or crystals |
| storage temp. room temp | storage temp. 15-25°C | storage temp. - | storage temp. 2-8°C |
Still not finding the right product?
Explore all of our products under RockPhos
Lagerklasse
11 - Combustible Solids
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.


