biological source
mouse
recombinant
expressed in NSO cells
assay
≥90% (SDS-PAGE)
form
lyophilized powder
mol wt
apparent mol wt 85-90 kDa by SDS-PAGE (reducing), calculated mol wt ~58 kDa
storage condition
avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)
impurities
endotoxin, tested
UniProt accession no.
storage temp.
−20°C
Gene Information
mouse ... Angptl1(72713)
Physical form
Lyophilized from a 0.2 μm filtered solution in 50 mM Tris citrate and 0.5 M sodium chloride, pH 6.5, containing 1.25 mg bovine serum albumin.
Analysis Note
The biological activity is measured by its ability to bind immobilized Tie-2/Fc in ELISA.
Lagerklasse
11 - Combustible Solids
Was ist los?
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.