Fortfahren mit
A2549
Tape, autoclave indicator, Sigmaware™
impregnated to show words Autoclaved
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
NACRES:
NB.35
UNSPSC Code:
55121611
General description
  • Tape is impregnated to show words Autoclaved after 15 minutes of exposure at 250°F (121°C) in a steam autoclave
  • Tape is supplied in 500 in. (1,270 cm) roll on a 1 in. core (2.54 cm)
  • Tape Dimensions: W 3/4 in. (1.91 cm)
  • Product contains lead which under normal use, does not present a health hazard or safety risk
  • Product contains latex (dry natural rubber)
Legal Information
Sigmaware is a trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)