Direkt zum Inhalt
Merck

Fortfahren mit

AV30009

Anti-FLJ14768 antibody produced in rabbit

IgG fraction of antiserum

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis
100 μL
Versand innerhalb von 5 Werktagen. (Bei Bestellungen außerhalb der USA rechnen Sie bitte zusätzlich 1-2 Wochen für die Lieferung ein)
€ 401,00

Über diesen Artikel

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
2

€ 401,00


Versand innerhalb von 5 Werktagen. (Bei Bestellungen außerhalb der USA rechnen Sie bitte zusätzlich 1-2 Wochen für die Lieferung ein)

​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

52 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FLJ14768(84922)

Immunogen

Synthetic peptide directed towards the C terminal region of human FLJ14768

Application

Anti-FLJ14768 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25 μg/ml. For immunohistochemistry of paraffin-embedded tissue sections, a concentration of 4-8 μg/ml is suitable.

Biochem/physiol Actions

FLJ14768 or Flt3-interacting zinc-finger protein Fiz1 interacts with Flt3 receptor tyrosine kinases to regulate hematopoietic and lymphoid cells. It acts as a repressor of NRL (neural retina leucine zipper) transcription factor that regulates several rod-specific genes including rhodopsin.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: HCGALYAALAALEEHRRVSHGEGGGEEAAAAAREREPASGEPPSGSGRGK

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
SAB2104520HPA074026SAB1407994
description

IgG fraction of antiserum

description

affinity isolated antibody

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

description

purified immunoglobulin, buffered aqueous solution

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

human

species reactivity

human, bovine, pig

species reactivity

human

species reactivity

human

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous glycerol solution

form

buffered aqueous solution


Still not finding the right product?

Probieren Sie unser Produkt-Auswahlhilfe, um Ihre Auswahl einzugrenzen


Lagerklasse

10 - Combustible liquids

Was ist los?

WGK 3

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen