Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
NACRES:
NA.81
UNSPSC Code:
12352207
eCl@ss:
32150313
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltentechnique(s)
cell culture | mammalian: suitable
shipped in
ambient
storage temp.
room temp
Application
Primary Human Umbilical Vein Endothelial Cells (HUVEC) are isolated from the vein of the umbilical cord. They are commonly used for physiological and pharmacological investigations, such as macromolecule transport, blood coagulation, angiogenesis, and fibrinolysis. HUVEC are isolated from single donors in Endothelial Cell Growth Medium 2, which is free of ECGS and contains additional IGF and VEGF, and provided in a proliferating format. (Human Umbilical Artery Endothelial Cells (HUAEC) from the same donor are available on request.)
Preparation Note
Click here for more information.
Analysis Note
Rigid quality control tests are performed for each lot of Endothelial Cells from large vessels. They are tested for cell morphology, adherence rate, and cell viability. Flow cytometric analyses for cell-type specific markers, e.g. von Willebrand Factor (vWF) and CD31, as well as Dil-Ac-LDL uptake assays are also carried out for each lot. Growth performance is tested through multiple passages up to 15 population doublings (PD) under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).
Other Notes
Recommended Plating Density: 5000 - 10000 cells per cm2Passage After Thawing: P1Tested Markers: von Willebrand Factor (vWF) positive, CD31 positive, Dil-Ac-LDL uptake positiveGuaranteed population doublings: >15
Recommended Primary Cell Culture Media:Link
Disclaimer
Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
Still not finding the right product?
Explore all of our products under Human Umbilical Vein Endothelial Cells 2 (HUVEC 2)
Lagerklasse
12 - Non Combustible Liquids
WGK
WGK 2
Flammpunkt (°F)
No data available
Flammpunkt (°C)
No data available
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.