Direkt zum Inhalt
Merck

Fortfahren mit

F3-87-8-1 Mouse x Mouse Hybridoma, ECACC 89062108

no DNA, Reactivity: Anti-CNS specific membrane glycoprotein, human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 89062108

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
41106514
eCl@ss:
32160702
Biological source:
Mouse x Mouse Hybridoma
Isotype:
IgG1
Technique(s):
cell culture | mammalian: suitable
Myeloma:
NS1/1
Reactivity:
Anti-CNS specific membrane glycoprotein, human
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


Produktname

F3-87-8-1, no DNA, Reactivity: Anti-CNS specific membrane glycoprotein, human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 89062108

biological source

Mouse x Mouse Hybridoma

packaging

pkg of vial of cells 89062108-1VL, tube of 5 μg 89062108-DNA-5UG

manufacturer/tradename

ECACC

ImmunoDon

BALB/c mouse spleen

Myeloma

NS1/1

Reactivity

Anti-CNS specific membrane glycoprotein, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Immunogen

Human brain glycoproteins depleted of the Thy-1 antigen

Biochem/physiol Actions

Mouse NS1/1 x Mouse BALB/c spleen
The antibody is directed against a brain specific determinant present on a glycoprotein with a molecular weight of 130kDa, (CNS gp130). The antibody interacts with the F3-87-8-1 antigen in non human mammalian species.

Preparation Note

Maintain cultures between 2-5x100,000 cells/ml; 5% CO2; 37°C. Cells die at 5,000,000 cells/ml. Require mouse macrophage feeders upon resuscitation
RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). Genomic DNA is not available for this cell line. Use of this product by commercial organizations may require a license agreement. Commercial organizations will be contacted by cellscreening@merckgroup.com to provide further information after the order request is submitted. Any other use of this cell line is prohibited without the express written permission of UKHSA. Inquiries regarding any other authorized use of this cell line may be directed to culturecollections@ukhsa.gov.uk.

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
CB_13082222CB_84112011CB_85062136
vibrant-m

F3-87-8-1

vibrant-m

OX-108

vibrant-m

OX-18

vibrant-m

H81-208.22.6

description

no DNA, Reactivity: Anti-CNS specific membrane glycoprotein, human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 89062108

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

84112011, Reactivity: Anti-MHC class I, rat, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

Reactivity: Anti-I-Ek, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 85062136, no DNA

storage temp.

−196°C

storage temp.

−196°C

storage temp.

-

storage temp.

-

ImmunoDon

BALB/c mouse spleen

ImmunoDon

BALB/c mouse spleen

ImmunoDon

-

ImmunoDon

-

packaging

tube of 5 μg 89062108-DNA-5UG, pkg of vial of cells 89062108-1VL

packaging

tube of 5 μg 13082222-DNA-5UG, pkg of vial of cells 13082222-1VL

packaging

pkg of vial of cells 84112011-1VL, tube of 5 μg 84112011-DNA-5UG

packaging

tube of 5 μg 85062136-DNA-5UG, pkg of vial of cells 85062136-1VL

manufacturer/tradename

ECACC

manufacturer/tradename

ECACC

manufacturer/tradename

ECACC

manufacturer/tradename

ECACC

Myeloma

NS1/1

Myeloma

NS1

Myeloma

-

Myeloma

-

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable




Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen



Questions

Reviews

No rating value

Active Filters