Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
biological source
gerbil connective tissue
manufacturer/tradename
ECACC
packaging
tube of 5 μg 96020931-DNA-5UG, pkg of vial of cells 96020931-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Fibroblast
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Biochem/physiol Actions
Preparation Note
Other Notes
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 85020204, human lung (fetal), Fibroblast | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 86041809, human nerve, Fibroblast/neuroblast |
| biological source gerbil connective tissue | biological source buffalo lung | biological source human lung (fetal) | biological source human nerve |
| morphology Fibroblast | morphology Fibroblast | morphology Fibroblast | morphology Fibroblast/neuroblast |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
| karyotype Not specified | karyotype 2n = 60 | karyotype 2n = 46, diploid | karyotype 2n = 46 |
| relevant disease(s) cancer | relevant disease(s) - | relevant disease(s) - | relevant disease(s) metastasis |
| manufacturer/tradename ECACC | manufacturer/tradename ECACC | manufacturer/tradename ECACC | manufacturer/tradename ECACC |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?