Fortfahren mit
E5514
Vertikal-Elektrophoreseeinheit höhenverstellbar, Einzelgel
gel W × H 16.5 cm × 14.5-28 cm, Not CE-approved
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
UNSPSC Code:
12352200
eCl@ss:
32150319
grade
electrophoresis grade
availability
not available in EU
gel W × H
16.5 cm × 14.5-28 cm
General description
  • Made from high quality acrylic with platinum electrodes
  • Long lasting silicone sponge gasket on upper reservoir for leak-free operation without grease
  • Safety covers with attached safety-tip power leads
  • Designed for use with unique gel-casting gasket; heavy-duty casting clamps compress gasket between glass plates, sealing without grease
For Slab Gel Electrophoresis:
  • Designed to use as little as 200 mL of buffer, and as little as 5 μg of sample per well
  • Use with gel casting accessories which offer the ability to cast one or two gels in a single assembly
  • Use in standard slab gel electrophoresis or in the second dimension of 2D electrophoresis
Each unit comes with base unit, upper and lower reservoirs with safety covers and attached power leads, and two gel-mounting spring clamps.
Slab and tube gel accessories are not included; order from the accessories list.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)