Fortfahren mit
HS120104
Pearl™ Pipette
adjustable, volume 100-1000 μL, blue, single-channel
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
1 ea
Warenkorb auf Verfügbarkeit prüfen
€ 190,00
Über diesen Artikel
NACRES:
NB.01
UNSPSC Code:
41121500
eCl@ss:
32070102
Material:
blue , white
€ 190,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
blue , white
feature
adjustable, single-channel
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120104
volume measuring range
100-1000 μL
General description
The Pearl is a single-channel pipette offering adjustable volume and precision liquid handling. The sleek, ergonomic design and reduced plunger pressure help reduce the chance of Repetitive Stress Injury (RSI), as the pipette requires 2/3 the operation pressure of most standard pipettes. The Pearl is tested in accordance with standards and a calibration certificate is supplied. Manufactured in accordance with ISO08655 and GLP regulations.
Features and Benefits
  • Internal metal parts for increased accuracy and calibration stability, and long life
  • Ergonomic Slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
  • Easy volume adjustment with locking tab to prevent accidental change
  • Separate tip ejector
  • Large digit volume display
  • Color coded and labeled with volume range
  • Both top and side have colored coded components for quick visual confirmation
  • CE certified and Manufactured in accordance with ISO08655 and GLP regulations
  • Autoclavable
Legal Information
Pearl is a trademark of Specac, Inc.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
HS120101HS120105HS120098
description
adjustable, volume 100-1000 μL, blue, single-channel
description
adjustable, volume 10-100 μL, blue, single-channel
description
adjustable, volume 500-5000 μL, pink, single-channel
description
adjustable, volume 0.5-10 μL, light blue, single-channel
material
white , blue
material
white , blue
material
pink , white
material
light blue , white
volume measuring range
100-1000 μL
volume measuring range
10-100 μL
volume measuring range
500-5000 μL
volume measuring range
0.5-10 μL
feature
adjustable, single-channel
feature
adjustable, single-channel
feature
adjustable, single-channel
feature
single-channel, adjustable
manufacturer/tradename
Heathrow Scientific HS120104
manufacturer/tradename
Heathrow Scientific HS120101
manufacturer/tradename
Heathrow Scientific HS120105
manufacturer/tradename
Heathrow Scientific HS120098
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
Still not finding the right product?
Explore all of our products under Pearl™ Pipette
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)