Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
10 ea | Warenkorb auf Verfügbarkeit prüfen | € 87,90 |
Über diesen Artikel
€ 87,90
material
(with Pivot ring), octagonal , round white PTFE stir bar(s)
packaging
pack of 10 ea
manufacturer/tradename
Heathrow Scientific 120551
L × diam.
30 mm × 8 mm
General description
Features and Benefits
- Magnetic bar is made from ALNICO, one of the strongest permanent magnets available
- The sintered manufacturing process reduces the chance of decoupling (spin outs), which weaker stir bars often suffer from
- Encapsulation in PTFE protects both magnet and what it is stirring
- Chemically inert and easy to clean
- Can withstand temperatures up to 250°C, making them suitable for use with both unheated and heated stirrers
- Low friction octagon shape with a center pivot ring helps create a strong mixing vortex, even with more viscous solutions
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description L × diam. 30 mm × 8 mm | description L × diam. 20 mm × 8 mm | description L × diam. 35 mm × 9 mm | description L × diam. 28 mm × 8 mm |
| manufacturer/tradename Heathrow Scientific 120551 | manufacturer/tradename Heathrow Scientific 120548 | manufacturer/tradename Heathrow Scientific 120552 | manufacturer/tradename Heathrow Scientific 120550 |
| material round white PTFE stir bar(s), (with Pivot ring), octagonal | material octagonal , (with Pivot ring), round white PTFE stir bar(s) | material (with Pivot ring), octagonal , round white PTFE stir bar(s) | material (with Pivot ring), octagonal , round white PTFE stir bar(s) |
| packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 10 ea |
| L × diam. 30 mm × 8 mm | L × diam. 20 mm × 8 mm | L × diam. 35 mm × 9 mm | L × diam. 28 mm × 8 mm |
Still not finding the right product?
Explore all of our products under Magnetic Stir Bar
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?