Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
square black polystyrene
size
small
packaging
pk of 500 ea
manufacturer/tradename
Heathrow Scientific HS1423A
L × W × H
(46 mm) × 46 mm × 8 mm
General description
Features and Benefits
- biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
- temperature range -10 to 70C
- disposable
- shallow with a wide, flat bottom to resist tipping
- rounded corners simplify transfer
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description Small, square black polystyrene, pk of 500 ea | description Small, square black polystyrene, Anti-static, pk of 500 ea | description Medium, black polystyrene (square), Anti-static, pk of 500 ea | description size small, square white polystyrene |
| manufacturer/tradename Heathrow Scientific HS1423A | manufacturer/tradename Heathrow Scientific HS1423AA | manufacturer/tradename Heathrow Scientific HS1423BB | manufacturer/tradename Heathrow Scientific HS1420A |
| material square black polystyrene | material square black polystyrene | material black polystyrene (square) | material square white polystyrene |
| packaging pk of 500 ea | packaging pk of 500 ea | packaging pk of 500 ea | packaging pack of 500 ea |
| size small | size small | size medium | size small |
| L × W × H (46 mm) × 46 mm × 8 mm | L × W × H (46 mm) × 46 mm × 8 mm | L × W × H (85 mm) × 84 mm × 24 mm | L × W × H 46 mm × 46 mm × 8 mm |
Still not finding the right product?
Explore all of our products under Weigh Boat
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?