Fortfahren mit
HS24312B
Peg Tube Rack
to hold, 96 tube(s) 10-13 mm tubes, blue
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
2 ea
Warenkorb auf Verfügbarkeit prüfen
€ 36,60
Über diesen Artikel
NACRES:
NB.17
UNSPSC Code:
41122800
Manufacturer/tradename:
Heathrow Scientific HS24312B
Feature:
to hold 96 tube(s) 10-13 mm tubes
Material:
blue , fiberglass , polypropylene
Packaging:
pack of 2 ea
€ 36,60
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
blue , fiberglass , polypropylene
feature
to hold 96 tube(s) 10-13 mm tubes
packaging
pack of 2 ea
manufacturer/tradename
Heathrow Scientific HS24312B
L × W × H
82 mm × 126 mm × 69 mm
General description
The Peg Tube Rack is strong, tough, and versatile for bench or tube storage. It is constructed of fiberglass-reinforced polypropylene, which is ideal for bench work or tube storage.
Features and Benefits
  • Constructed of fiberglass-reinforced polypropylene
  • Spacious peg design allows easy viewing of tube contents
  • Use as a drying rack for glass plates or upside-down tubes
  • Pegs remain straight even after repeated autoclaving
  • Embossed alphanumeric identification for quick location and retrieval of tubes
  • Unique tear drop well design allows for better retention of tubes
  • Autoclavable
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
HS24312AHS24311BHS24311A
Peg Tube Rack to hold, 96 tube(s) 10-13 mm tubes, blue
HS24312B
Peg Tube Rack
Peg Tube Rack to hold, 96 tube(s) 10-13 mm tubes, white
HS24312A
Peg Tube Rack
Peg Tube Rack to hold, 50 tube(s) 14-17 mm tubes, blue
HS24311B
Peg Tube Rack
Peg Tube Rack to hold, 50 tube(s) 14-17 mm tubes, white
HS24311A
Peg Tube Rack
description
to hold, 96 tube(s) 10-13 mm tubes, blue
description
to hold, 96 tube(s) 10-13 mm tubes, white
description
to hold, 50 tube(s) 14-17 mm tubes, blue
description
to hold, 50 tube(s) 14-17 mm tubes, white
manufacturer/tradename
Heathrow Scientific HS24312B
manufacturer/tradename
Heathrow Scientific HS24312A
manufacturer/tradename
Heathrow Scientific HS24311B
manufacturer/tradename
Heathrow Scientific HS24311A
feature
to hold 96 tube(s) 10-13 mm tubes
feature
to hold 96 tube(s) 10-13 mm tubes
feature
to hold 50 tube(s) 14-17 mm tubes
feature
to hold 50 tube(s) 14-17 mm tubes
material
blue , fiberglass , polypropylene
material
fiberglass , polypropylene , white
material
blue , fiberglass , polypropylene (reinforced)
material
polypropylene (reinforced), fiberglass , white
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 2 ea
L × W × H
82 mm × 126 mm × 69 mm
L × W × H
82 mm × 126 mm × 69 mm
L × W × H
-
L × W × H
188 mm × 104 mm × 69 mm
Still not finding the right product?
Explore all of our products under Peg Tube Rack
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)