Fortfahren mit
Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
AC/DC input
, universal plug set, 220 V / 240 V
feature
electrode InLab® Pure Pro-ISM
packaging
pack of 1 kit
manufacturer/tradename
Mettler-Toledo 30671548
concentration measuring range
-2.000-20.000 pX, resolution: 3 or 4 digits (ion), 0-999,999 mg/L (ion)
mV measuring range
-2,000.000-2,000.000 mV, resolution: 1; 0.1
pH measuring range
accuracy: ±0.002, resolution: 0.1, resolution: 0.001, resolution: 0.01
temperature measuring range
-30-130 °C, resolution: 0.1 °C
General description
Large color touchscreen provides direct measurement and easy data entry. Instructions guide users through the sensor calibration process.
Tracks your Records
The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
Tough Enough
Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
pH Sensor for Pure Water
InLab Pure-Pro ISM is a pH/temperature sensor designed for accurate and stable pH measurements in pure water. The brige electrolyte, sleeve junction, and low-resistance glass membrane support accurate measurements in ion-poor media.
Electrode Arm for Efficient Operation
The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy. Dedicated positions for cleaning and storage support correct sensor handling. Vertical and horizontal movements facilitate precise positioning, even with one hand.
Protect Critical Settings
A two-level user management protects the most important settings. Operators retain the flexibility to individually set preferences like language and screen brightness.
Display Adapts to Your Needs
View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
Connectors: BNC; RCA / Cinch; 2 mm Reference; USB A (x2); USB B; Mini DIN; (for stirrer)
Application
Features and Benefits
- Large color touchscreen provides direct measurement and easy data entry.
- The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
- Smart design and cover keep the instrument safe from dust and spills
- The pH/temperature sensor designed for accurate and stable pH measurements in pure water
- Electrode arm for efficient operation
- Protect critical settings Protected with two-level user management
- Display adapts to your needs to maximize readability
Other Notes
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description electrode, InLab® Pure Pro-ISM | description electrode, InLab® Science Pro-ISM | description electrode, InLab® Routine Pro-ISM | description meter and electrode arm only |
| AC/DC input 220 V / 240 V, , universal plug set | AC/DC input , universal plug set, 220 V / 240 V | AC/DC input 220 V / 240 V, , universal plug set | AC/DC input 220 V / 240 V, universal plug set |
| packaging pack of 1 kit | packaging pack of 1 kit | packaging pack of 1 kit | packaging pack of 1 kit |
| feature electrode InLab® Pure Pro-ISM | feature electrode InLab® Science Pro-ISM | feature electrode InLab® Routine Pro-ISM | feature - |
| pH measuring range resolution: 0.01, resolution: 0.001, accuracy: ±0.002, resolution: 0.1 | pH measuring range resolution: 0.001, resolution: 0.01, accuracy: ±0.002, resolution: 0.1 | pH measuring range resolution: 0.01, resolution: 0.001, accuracy: ±0.002, resolution: 0.1 | pH measuring range accuracy: ±0.002, resolution: 0.1, resolution: 0.01, resolution: 0.001 |
| temperature measuring range -30-130 °C, resolution: 0.1 °C | temperature measuring range -30-130 °C, resolution: 0.1 °C | temperature measuring range -30-130 °C, resolution: 0.1 °C | temperature measuring range -30-130 °C, resolution: 0.1 °C |
| manufacturer/tradename Mettler-Toledo 30671548 | manufacturer/tradename Mettler-Toledo 30671547 | manufacturer/tradename Mettler-Toledo 30671546 | manufacturer/tradename Mettler-Toledo 30671552 |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.



