Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
NACRES:
NB.15
UNSPSC Code:
24111500
Manufacturer/tradename:
MTC™ Bio A8001R
Feature:
autoclavable, label with large marking area and sterilization indicator
Material:
high-density polyethylene
Packaging:
pack of 100 ea
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
high-density polyethylene
feature
autoclavable, label with large marking area and sterilization indicator
packaging
pack of 100 ea
manufacturer/tradename
MTC™ Bio A8001R
W × L
27 in. (64 cm) × 35 in. (89 cm)
capacity
12 gal (45 L)
thickness
2.25 mil
color
red
General description
The exclusive drawstring loops on BowTie™ Biohazard Bagsprovide instant-cinch closure on your waste materials and a handle to easilyremove bags from their bins. They allow venting during sterilization andprovide an easy, safe way to remove bags from a hot autoclave. Made from 2.25 mil thick HDPE these bags ares suitable for autoclaving in 121°C cycles and are printed with a number of identifyingmarkers such as large biohazard symbols and temperature limit warnings. Theyare also outfitted with a large marking area and a sterilization patch thatchanges colour after autoclaving. BowTie™ Bags fit their respectivebiohazard bins perfectly.
Features and Benefits
- Cinch closure, no wires needed
- Colour changing sterilization indicator
- Biohazardsymbols and temperature limit warnings
- Allow venting during sterilization
- Safe& easiy removal from bin or autoclave
- 2.25mil thick HDPE
Legal Information
BowTie is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Still not finding the right product?
Explore all of our products under MTC™ Bio BowTie™ Drawstring Biohazard Bags
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?