Fortfahren mit
MTCA8403SK
MTC™ Bio BenchBin™ starter Set
includes 3L BenchBin™ biohazard bin & 400 red autoclave/biohazard bags
Synonym(e):
MTC™ Bio BenchBin™ starter Set, bio waste bin, MTC Bio A8403-SK, disposal bin, waste bin
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
24111500
NACRES:
NB.43
eCl@ss:
32049090
Manufacturer/tradename:
MTC™ Bio A8403-SK
Material:
red (with black Biohazard warning)
Packaging:
pack of 1 ea
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
red (with black Biohazard warning)
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio A8403-SK
capacity
3 L
General description
BenchBin™ is a personal benchtop container for biohazard and contaminated waste. Compact size fits perfectly anywhere and safely disposes up to tips, tubes, gloves, etc. Features easy push open/close lid that is perfect for quick disposal of biological waste. The cover is easily removed from the inner basin for quick emptying of contents. Ideal size for use with red imprinted autoclavable biohazard bags. BenchBin Starter kit includes one BenchBin and 400 red biohazard bags.
Features and Benefits
  • Compact size
  • Safely disposes tips, tubes, gloves
  • Easy open lid
  • Research or further manufacturing use only, not for food or drug use.
Legal Information
BenchBin is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
MTCA8403BMTCA8006BMTCA8002B
description
includes 3L BenchBin™ biohazard bin & 400 red autoclave/biohazard bags
description
capacity 3 L, with spring loaded lid, pack of 1 ea
description
capacity 30 L (50 gal), W × L × H 6 in. (41 cm) × 16 in. (41 cm) × 24 in. (60 cm), pack of 1 ea
description
capacity 26 gal (100 L), W × L 19 in. (48 cm) × 19 in. (48 cm)
manufacturer/tradename
MTC™ Bio A8403-SK
manufacturer/tradename
MTC™ Bio A8403B
manufacturer/tradename
MTC™ Bio A8006B
manufacturer/tradename
MTC™ Bio A8002B
material
red (with black Biohazard warning)
material
red (with black Hiohard warning)
material
red (with black Biohazard warning)
material
red (with black Biohazard warning)
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
capacity
3 L
capacity
3 L
capacity
30 L (50 gal)
capacity
26 gal (100 L)
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)