Fortfahren mit
MTCC2231W
MTC™ Bio ScrewSeal™ Microcentrifuge Tubes
self-standing, clear, capacity 2.0 mL, pkg of 500 ea (individually wrapped)
Synonym(e):
conical tube, plastic tube, MTC Bio C2231-W, sample collection tube, sample receptacle, sample tube
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
NACRES:
NB.15
UNSPSC Code:
41121800
eCl@ss:
32039090
Capacity:
2.0 mL
Material:
blue cap, clear , high-density polyethylene cap, polypropylene , screw top cap, self-standing
Sterility:
sterile
Packaging:
pkg of 500 ea (individually wrapped)
Parameter:
22,000 × g speed
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
blue cap, clear , high-density polyethylene cap, polypropylene , screw top cap, self-standing
sterility
sterile
packaging
pkg of 500 ea (individually wrapped)
manufacturer/tradename
MTC™ Bio C2231-W
parameter
22,000 × g speed
capacity
2.0 mL
diam. × L
12.8 mm × 46 mm
General description
Screw-cap microtubes, 2.0mL, with O-ring, , molded graduations, caps attached, sterile, individually wrapped, 500/cs
ScrewSeal™ screw-cap microtubes are made from ultra-clear polypropylene, these screw cap microtubes include assembled caps with silicone O-rings that assure a leak-proof seal from -90°C to +121°C. Sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000×g.
Features and Benefits
  • Ultra-clear polypropylene
  • Molded or non-graduated options
  • Leak-proof O-ring seal
  • Light-blocking amber available
  • Sterile, RNase, DNase, Pyrogen free
Legal Information
MTC is a trademark of MTC Bio
ScrewSeal is a trademark of MTC Bio
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)