Direkt zum Inhalt
Merck

Fortfahren mit

MTCD309617

MTC™ Bio OptiWell™ Deep Well Plate

96 round wells, V-bottom wells, capacity 1.0 mL, non-sterile, case of 50 ea (10 bags of 5ea)

Synonym(e):

automated research plate, MTC Bio D3096-17, automated assay plate, lab automation plate, research automation tray, robotic research plate

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
41121800
NACRES:
NB.15
Material:
V-bottom wells, clear plate, clear wells, polypropylene , round wells
Size:
96 wells
Sterility:
non-sterile
Feature:
lid: no, skirt: no
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


material

V-bottom wells, clear plate, clear wells, polypropylene , round wells

sterility

non-sterile

feature

lid: no, skirt: no

packaging

case of 50 ea (10 bags of 5ea)

manufacturer/tradename

MTC™ Bio D3096-17

parameter

4,000 × g speed

capacity

1.0 mL

size

96 wells

well volume

1.0 mL

General description

OptiWell plates come in 24, 48, 96 or 384 format for virtually every research lab, HTS application and automated/robotic platform. These plates are useful for bacterial cultures, centrifugation (up to 4000 x g) and general sample storage. Plates are produced/packaged in a Class 100,000 clean room and are ANSI / SLAS 1-2004 compliant. Several plates are sterilized by gamma irradiation and all are certified non-pyrogenic, RNase/DNase free.

Features and Benefits

  • For automated research platforms / robotic equipment
  • Produced/packaged in a Class 100,000 clean room
  • ANSI / SLAS 1-2004 compliant
  • Minimal binding / liquid retention
  • Black printed alpha-numerics
  • Virgin, medical-grade polypropylene
  • Sterilized by gamma irradiation
  • Certified non-pyrogenic, RNase/DNase free

Legal Information

MTC is a trademark of MTC Bio
OptiWell is a trademark of MTC Bio

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
MTCD309635MTCD309625MTCD309621
description

96 round wells, V-bottom wells, capacity 1.0 mL, non-sterile, case of 50 ea (10 bags of 5ea)

description

96 round wells, U-bottom wells, capacity 2.0 mL, non-sterile, case of 50 ea (10 bags of 5ea)

description

96 square wells, U-bottom wells, capacity 1.2 mL, non-sterile, case of 50 ea (10 bags of 5ea)

description

96 round wells, U-bottom wells, capacity 1.1 mL, non-sterile, case of 50 ea (10 bags of 5ea)

manufacturer/tradename

MTC™ Bio D3096-17

manufacturer/tradename

MTC™ Bio D3096-35

manufacturer/tradename

MTC™ Bio D3096-25

manufacturer/tradename

MTC™ Bio D3096-21

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

feature

lid: no, skirt: no

feature

lid: no, skirt: no

feature

skirt: no, lid: no

feature

lid: no, skirt: no

material

clear plate, clear wells, round wells, V-bottom wells, polypropylene

material

U-bottom wells, clear wells, clear plate, polypropylene , round wells

material

polypropylene , U-bottom wells, clear wells, square wells, clear plate

material

clear wells, round wells, U-bottom wells, clear plate, polypropylene

size

96 wells

size

96 wells

size

96 wells

size

96 wells

packaging

case of 50 ea (10 bags of 5ea)

packaging

case of 50 ea (10 bags of 5ea)

packaging

case of 50 ea (10 bags of 5ea)

packaging

case of 50 ea (10 bags of 5ea)


Still not finding the right product?




Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen