Fortfahren mit
MTCD309625
MTC™ Bio OptiWell™ Deep Well Plate
96 square wells, U-bottom wells, capacity 1.2 mL, non-sterile, case of 50 ea (10 bags of 5ea)
Synonym(e):
automated research plate, MTC Bio D3096-25, automated assay plate, lab automation plate, research automation tray, robotic research plate
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41121800
NACRES:
NB.15
Material:
U-bottom wells, clear plate, clear wells, polypropylene , square wells
Size:
96 wells
Sterility:
non-sterile
Feature:
skirt: no, lid: no
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
U-bottom wells, clear plate, clear wells, polypropylene , square wells
sterility
non-sterile
feature
skirt: no, lid: no
packaging
case of 50 ea (10 bags of 5ea)
manufacturer/tradename
MTC™ Bio D3096-25
parameter
4,000 × g speed
capacity
1.2 mL
size
96 wells
well volume
1.2 mL
General description
OptiWell plates come in 24, 48, 96 or 384 format for virtually every research lab, HTS application and automated/robotic platform. These plates are useful for bacterial cultures, centrifugation (up to 4000 x g) and general sample storage. Plates are produced/packaged in a Class 100,000 clean room and are ANSI / SLAS 1-2004 compliant. Several plates are sterilized by gamma irradiation and all are certified non-pyrogenic, RNase/DNase free.
Features and Benefits
  • For automated research platforms / robotic equipment
  • Produced/packaged in a Class 100,000 clean room
  • ANSI / SLAS 1-2004 compliant
  • Minimal binding / liquid retention
  • Black printed alpha-numerics
  • Virgin, medical-grade polypropylene
  • Sterilized by gamma irradiation
  • Certified non-pyrogenic, RNase/DNase free
Legal Information
MTC is a trademark of MTC Bio
OptiWell is a trademark of MTC Bio
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
MTCD309627MTCD309625SMTCD309635
description
96 square wells, U-bottom wells, capacity 1.2 mL, non-sterile, case of 50 ea (10 bags of 5ea)
description
96 round wells, U-bottom wells, capacity 1.2 mL, non-sterile, case of 50 ea (10 bags of 5ea)
description
96 square wells, U-bottom wells, capacity 1.2 mL, sterile, case of 50 ea (10 bags of 5ea)
description
96 round wells, U-bottom wells, capacity 2.0 mL, non-sterile, case of 50 ea (10 bags of 5ea)
manufacturer/tradename
MTC™ Bio D3096-25
manufacturer/tradename
MTC™ Bio D3096-27
manufacturer/tradename
MTC™ Bio D3096-25S
manufacturer/tradename
MTC™ Bio D3096-35
sterility
non-sterile
sterility
non-sterile
sterility
sterile
sterility
non-sterile
feature
lid: no, skirt: no
feature
skirt: no, lid: no
feature
skirt: no, lid: no
feature
lid: no, skirt: no
material
clear plate, polypropylene , square wells, U-bottom wells, clear wells
material
clear wells, clear plate, polypropylene , round wells, U-bottom wells
material
polypropylene , clear plate, clear wells, U-bottom wells, square wells
material
polypropylene , clear wells, clear plate, U-bottom wells, round wells
size
96 wells
size
96 wells
size
96 wells
size
96 wells
packaging
case of 50 ea (10 bags of 5ea)
packaging
case of 50 ea (10 bags of 5ea)
packaging
case of 50 ea (10 bags of 5ea)
packaging
case of 50 ea (10 bags of 5ea)
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)