Fortfahren mit
MTCF4060F
MTC™ Bio BioFlask™ Erlenmeyer Shaker Flask, Individually Wrapped
capacity 125 mL, polypropylene, flat bottom, cap, vented, sterile
Synonym(e):
culture flask, MTC Bio F4060-F, laboratory flask, plastic flask, shaker flask
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
NACRES:
NB.15
UNSPSC Code:
41122104
Capacity:
125 mL
Material:
flat bottom, polypropylene
Sterility:
sterile
Feature:
cap vented
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
flat bottom, polypropylene
sterility
sterile
feature
cap vented
packaging
pack of 24 ea (individually wrapped)
manufacturer/tradename
MTC™ Bio F4060-F
capacity
125 mL
General description
Erlenmeyer Shaker Flasks are made of polycarbonate or PETG. The polypropylene screw caps are vented with 0.2µm membranes to increase air transmission and enhance cell growth. The mouth size conforms with the international SBS standard, for quick, secure connection to bioreactor equipment. Polycarbonate flasks have baffled (ribbed) bottoms to provide increased oxygenation and mixing efficiency even at lower shaking speeds. Polycarbonate Flasks are autoclavable at 121°C and 15 psi. All flasks, regardless of size, are individually wrapped and gamma sterilized to SAL 10-6. They are free of RNase, DNase, pyrogen, endotoxins and human DNA.
Flask, Erlenmeyer, 125ml, PETG, flat bottom w / vented screw cap, sterile, indiv wrapped, 24 / CS
Features and Benefits
  • Vented screw caps
  • Flat or baffled bottoms
  • Gamma sterile (SAL 10-6)
  • Pyrogen free and RNase/DNase free
  • Individually wrapped
Legal Information
BioFlask is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)