Merck
MTCH680050
MTC™ Bio Halo™ Lightweight Single Channel Pipettor
capacity 5-50 μL
Synonym(e):
pipette, MTC Bio H6800-50, handheld liquid transfer device, liquid transfer device, mechanical pipettor
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
NACRES:
NB.15
UNSPSC Code:
41121500
eCl@ss:
32150313
Packaging:
pack of 1 ea
Feature:
adjustable, manual, no. of channels 1
Manufacturer/tradename:
MTC™ Bio H6800-50
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
feature
adjustable, manual, no. of channels 1
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio H6800-50
capacity
5-50 μL
General description
As one of the lightest pipettors ever introduced, the Halo™ Pipettor dramatically reduces the risk of repetitive stress injury and the general shoulder, arm, and hand fatigue associated with daily pipetting. Reduced actuation forces andan ergonomic design make Halo™ Pipettors feel balanced and natural in hand. Featuring a four-digit micrometer display for precise volume selection and a click-based adjustment wheel with DriftGuard protection, the Halo™ Pipettor is perfect for applications that demand exceptional accuracy. With a recessed ejector button, and cushioned, shock-absorbing stop, the ergonomics of tip ejection have been re-imagined to relieve stress on the thumb and joints.
Halo™ Single Channel Pipettor, lightweight, 0.5-5mL
Features and Benefits
  • 10-40% lighter than leading competitors
  • 4 Digit micrometer with DriftGuard protection
  • Exclusive 1250µL model for extended length tips
  • User-enabled calibration port
  • Compatible with universal tips
Legal Information
Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)