Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
polystyrene
sterility
sterile
packaging
case of 1000 ea (50 peel packs of 20ea)
manufacturer/tradename
MTC™ Bio M4027
capacity
10 μL
General description
Flexible Polypropylene Loops - Make it easier to maneuver them within Petri dishes, test tubes, or other confined spaces, improving the researcher′s ability to collect and transfer samples efficiently. The flexibility allows for more precise sampling from cultures, especially when working with small or delicate colonies that could be easily damaged by more rigid instruments.
Rigid Polystyrene Loops & Needles - Allow for more precise streaking patterns on agar plates, which is essential for isolating individual colonies or performing quantitative assays. They are also better suited for handling viscous or dense samples, where flexibility might impede the ability to effectively collect and transfer the sample.
Features and Benefits
- Sterilized to SAL 10-6
- Easy-grip hexagonal handle design
- Flexible PP or Rigid PS available
- Individually wrapped or peel packs of 20
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description rigid loop, capacity 10 μL, polystyrene, sterile, case of 1000 ea (50 peel packs of 20ea) | description rigid loop, capacity 10 μL, polystyrene, sterile, case of 1000 ea (individually wrapped) | description flexible loop, capacity 1 μL, polypropylene, sterile, case of 1000 ea (individually wrapped) | description flexible loop, capacity 10 μL, polypropylene, sterile, case of 1000 ea (individually wrapped) |
| material polystyrene | material polystyrene | material polypropylene | material polypropylene |
| capacity 10 μL | capacity 10 μL | capacity 1 μL | capacity 10 μL |
| packaging case of 1000 ea (50 peel packs of 20ea) | packaging case of 1000 ea (individually wrapped) | packaging case of 1000 ea (individually wrapped) | packaging case of 1000 ea (individually wrapped) |
| manufacturer/tradename MTC™ Bio M4027 | manufacturer/tradename MTC™ Bio M4027-W | manufacturer/tradename MTC™ Bio M4015-W | manufacturer/tradename MTC™ Bio M4025-W |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
Explore all of our products under MTC™ Bio Inoculating Loops and Needles
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?